Skip to Content
Merck
All Photos(2)

Key Documents

810332C

Avanti

18:1 PE CF

Avanti Research - A Croda Brand 810332C

Synonym(s):

1,2-dioleoyl-sn-glycero-3-phosphoethanolamine-N-(carboxyfluorescein) (ammonium salt)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C62H94N3O14P
CAS Number:
Molecular Weight:
1136.40
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

Assay

>99% (TLC)

form

solution

packaging

pkg of 1 × 1 mL (810332C-1mg)
pkg of 5 × 1 mL (810332C-5mg)
pkg of 5 × 2 mL (810332C-10mg)

manufacturer/tradename

Avanti Research - A Croda Brand 810332C

concentration

1 mg/mL (810332C-10mg)
1 mg/mL (810332C-1mg)
1 mg/mL (810332C-5mg)

shipped in

dry ice

storage temp.

−20°C

General description

Phosphoethanolamine (PHOS) is a precursor to phosphatidylcholine and phosphatidylethanolamine. 18:1 PE CF is a green fluorescent tagged phosphatidylethanolamine.

Application

18:1 PE CF may be used:
  • to spike human serum
  • to prepare concentrated lipid mix, PC:PE:fluoPE to assess basal binding properties and PC:PE:fluoPE:PIP to assess specific binding properties of the protein to each phosphoinositides (PIP)
  • to prepare the large unilamellar vesicles (LUVs)
  • in the preparation of fluorescent phosphoinositide?containing liposomes
  • for fluorescent labeling of total lipids to the initial liposome formulation mixture
  • in the formulation of pegylated magneto-liposomes (MLs) and transferrin (Tf) embedding

Biochem/physiol Actions

Phosphatidylethanolamine controls the membrane fluidity in eukaryotic cells. Phosphoethanolamine (PHOS) has the ability to repress tumour growth both in vitro and in vivo. It possesses anticarcinogenic effects. It can also prevent the mRNA expression of cyclin D1 and VEGFR1 (vascular endothelial growth factor receptor 1).

Packaging

5 mL Amber Glass Screw Cap Vial (810332C-10mg)
5 mL Amber Glass Screw Cap Vial (810332C-1mg)
5 mL Amber Glass Screw Cap Vial (810332C-5mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 Oral - STOT SE 3

Target Organs

Central nervous system, Liver,Kidney

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Liposome Flotation Assays for Phosphoinositide-protein Interaction
Tronchere H and Boal F
Journal of Cell Science, 390(1), 1-13 (2015)
Protein-Lipid Interaction by Fluorescence (PLIF) to Characterize and Screen for Inhibitors of Protein-Phosphoinositide Interactions
Ceccato L, et al.
Current Protocols in Protein Science / Editorial Board, John E. Coligan ... [Et Al.], 89(1), 19-31 (2017)
NUTS and BOLTS: applications of fluorescence-detected sedimentation
Kroe RR and Laue TM
Analytical biochemistry, 390(1), 1-13 (2009)
Enhanced blood-brain barrier transmigration using a novel transferrin embedded fluorescent magneto-liposome nanoformulation
Ding H, et al.
Nanotechnology, 25(5), 055101-055101 (2014)
Phosphoethanolamine and the danger of unproven drugs
Ponde N, et al.
Ecancermedicalscience, 10(7), 3658-3667 (2016)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service