Skip to Content
Merck
All Photos(1)

Key Documents

700126P

Avanti

campesterol

Avanti Research - A Croda Brand 700126P, powder

Synonym(s):

campest-5-en-3β-ol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C28H48O
CAS Number:
Molecular Weight:
400.68
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 1 mg (700126P-1mg)
pkg of 1 × 10 mg (700126P-10mg)

manufacturer/tradename

Avanti Research - A Croda Brand 700126P

shipped in

dry ice

storage temp.

−20°C

SMILES string

OC1CCC2(C3C(C4C(C(CC4)C(CCC(C(C)C)C)C)(CC3)C)CC=C2C1)C

InChI

1S/C28H48O/c1-18(2)19(3)7-8-20(4)24-11-12-25-23-10-9-21-17-22(29)13-15-27(21,5)26(23)14-16-28(24,25)6/h9,18-20,22-26,29H,7-8,10-17H2,1-6H3

InChI key

SGNBVLSWZMBQTH-UHFFFAOYSA-N

General description

Campesterol is a plant sterol, which is an analog of cholesterol and a brassinosteroid (BR) precursor. The enzyme diminute/dwarf1 mediates the synthesis of campesterol from 24-methylenecholesterol. Campesterol possesses methyl group at C-24 position of the side chain in the cholesterol structure.

Application

Campesterol may be used as a plant sterol to test its effect on the transcriptional activation of liver X receptor α (LXRA) in breast cancer cells. It may also be used as a sterol standard for calibration curve generation in liquid chromatography multiple reaction monitoring (LC-MRM) analysis.

Biochem/physiol Actions

Campesterol reduces the levels of chylomicron, low-density lipoprotein (LDL) and very low-density lipoprotein (VLDL)-apoB100. A high ratio of campesterol to sitosterol enhances fiber elongation in cotton fibres.

Packaging

5 mL Amber Glass Screw Cap Vial (700126P-10mg)
5 mL Amber Glass Screw Cap Vial (700126P-1mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

also commonly purchased with this product

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Laura Calpe-Berdiel et al.
Atherosclerosis, 203(1), 18-31 (2008-08-12)
Plant sterols and stanols (phytosterols/phytostanols) are known to reduce serum low-density lipoprotein (LDL)-cholesterol level, and food products containing these plant compounds are widely used as a therapeutic dietary option to reduce plasma cholesterol and atherosclerotic risk. The cholesterol-lowering action of
Shasha Deng et al.
Science China. Life sciences, 59(2), 183-193 (2016-01-25)
Phytosterols play an important role in plant growth and development, including cell division, cell elongation, embryogenesis, cellulose biosynthesis, and cell wall formation. Cotton fiber, which undergoes synchronous cell elongation and a large amount of cellulose synthesis, is an ideal model
U Klahre et al.
The Plant cell, 10(10), 1677-1690 (1998-10-08)
We have identified the function of the Arabidopsis DIMINUTO/DWARF1 (DIM/DWF1) gene by analyzing the dim mutant, a severe dwarf with greatly reduced fertility. Both the mutant phenotype and gene expression could be rescued by the addition of exogenous brassinolide. Analysis
Michael Igel et al.
Journal of lipid research, 44(3), 533-538 (2003-02-04)
The recent identification of the aberrant transport proteins ABCG5 and ABCG8 resulting in sitosterolemia suggests that intestinal uptake of cholesterol is an unselective process, and that discrimination between cholesterol and plant sterols takes place at the level of sterol efflux
Samantha A Hutchinson et al.
International journal of molecular sciences, 20(13) (2019-07-05)
Low fruit and vegetable consumption and high saturated fat consumption causes elevated circulating cholesterol and are breast cancer risk factors. During cholesterol metabolism, oxysterols form that bind and activate the liver X receptors (LXRs). Oxysterols halt breast cancer cell proliferation

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service