Skip to Content
Merck
All Photos(2)

Key Documents

700055P

Avanti

5α,6β-dihydroxycholestanol-d7

Avanti Research - A Croda Brand

Synonym(s):

3β,5α,6β-trihydroxycholestane (d7)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H41D7O3
CAS Number:
Molecular Weight:
427.71
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

description

cholestane-3β,5α,6β-triol-d7

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 1 mg (700055P-1mg)
pkg of 1 × 5 mg (700055P-5mg)

manufacturer/tradename

Avanti Research - A Croda Brand

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@@]12[C@]([C@](CC[C@H](O)C3)(C)[C@@]3(O)[C@H](O)C2)([H])CC[C@@]4(C)[C@@]1([H])CC[C@]4([H])[C@]([H])(C)CCCC([2H])(C([2H])([2H])[2H])C([2H])([2H])[2H]

InChI key

YMMFNKXZULYSOQ-UCHKBJBUSA-N

General description

5α,6β-dihydroxycholestanol is a structural derivative of cholesterol and is an abundant oxysterols. 5α,6β-dihydroxycholestanol-d7 is a deuterated form of 5α,6β-dihydroxycholestanol.

Application

5α,6β-dihydroxycholestanol-d7 has been used as an analyte in liquid chromatography with tandem mass spectrometry (LC-MS-MS) analysis based multiple reaction monitoring (MRM) studies and as an internal standard in tandem mass spectrometry (MS/MS) for analysis of plasma samples.

Biochem/physiol Actions

5α,6β-dihydroxycholestanol (5α,6β-diOH-Chol) or cholestane-3β,5α,6β-triol (C-triol) levels are elevated along with 7-ketocholesterol in Niemann-Pick type C (NP-C) disease and is a key biomarkers for detection. It has ,neuroprotective functionality and may elicit protection against neuronal injury during ischemia.

Packaging

5 mL Amber Glass Screw Cap Vial (700055P-1mg)
5 mL Amber Glass Screw Cap Vial (700055P-5mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

also commonly purchased with this product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

No data available

Flash Point(C)

No data available


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sara Boenzi et al.
Journal of lipid research, 57(3), 361-367 (2016-01-07)
Oxysterols are intermediates of cholesterol metabolism and are generated from cholesterol via either enzymatic or nonenzymatic pathways under oxidative stress conditions. Cholestan-3β,5α,6β-triol (C-triol) and 7-ketocholesterol (7-KC) have been proposed as new biomarkers for the diagnosis of Niemann-Pick type C (NP-C)
Haiyan Hu et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 34(34), 11426-11438 (2014-08-22)
Overstimulation of NMDA-type glutamate receptors is believed to be responsible for neuronal death of the CNS in various disorders, including cerebral and spinal cord ischemia. However, the intrinsic and physiological mechanisms of modulation of these receptors are essentially unknown. Here
Glynis Klinke et al.
Clinical biochemistry, 48(9), 596-602 (2015-03-31)
Niemann-Pick type C (NP-C) is a rare progressive neurodegenerative lipid storage disorder with heterogeneous clinical presentation and challenging diagnostic procedures. Recently oxysterols have been reported to be specific biomarkers for NP-C but knowledge on the intra-individual variation and on reference
Ren Sheng et al.
Nature communications, 5, 4393-4393 (2014-07-16)
Wnt proteins control diverse biological processes through β-catenin-dependent canonical signalling and β-catenin-independent non-canonical signalling. The mechanisms by which these signalling pathways are differentially triggered and controlled are not fully understood. Dishevelled (Dvl) is a scaffold protein that serves as the
M Voorink-Moret et al.
Molecular genetics and metabolism, 123(2), 76-84 (2018-01-02)
In patients suspected of a lipid storage disorder (sphingolipidoses, lipidoses), confirmation of the diagnosis relies predominantly on the measurement of specific enzymatic activities and genetic studies. New UPLC-MS/MS methods have been developed to measure lysosphingolipids and oxysterols, which, combined with

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service