Key Documents
Z615684
Julabo® bath test tube racks
to fit 90 test tubes, for water baths, diam. 12-13 mm, polypropylene
Synonym(s):
tube holder
About This Item
UNSPSC Code:
41122804
NACRES:
NB.77
material
polypropylene
feature
Holds 90 x 12-13 mm tubes
packaging
pack of 1 ea
manufacturer/tradename
Julabo 8970381
parameter
+80 °C temp. limit
diam.
12-13 mm
General description
Polypropylene tray and test tube inserts for 90 test tubes with a 12/13 mm diameter, for use up to +80 °C. When tempering solvent-containing or acidic samples above 60 °C, it is recommended that stainless steel test tube inserts are used, as the plastic can be attacked.
Legal Information
Julabo is a registered trademark of Julabo USA, Inc.