Skip to Content
Merck
All Photos(1)

Key Documents

W508500

Sigma-Aldrich

Dimethyl phthalate

≥99%

Synonym(s):

DMP

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H4-1,2-(CO2CH3)2
CAS Number:
Molecular Weight:
194.18
Beilstein:
1911460
EC Number:
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
NACRES:
NA.21
Organoleptic:
odorless
grade:
Fragrance grade
biological source:
synthetic
Agency:
follows IFRA guidelines
food allergen:
no known allergens

biological source

synthetic

Quality Level

grade

Fragrance grade

Agency

follows IFRA guidelines

reg. compliance

EU Regulation 1223/2009

Assay

≥99%

form

liquid

autoignition temp.

1033 °F

refractive index

n20/D 1.515 (lit.)

bp

282 °C (lit.)

mp

2 °C (lit.)

density

1.19 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

no known allergens

Organoleptic

odorless

SMILES string

COC(=O)c1ccccc1C(=O)OC

InChI

1S/C10H10O4/c1-13-9(11)7-5-3-4-6-8(7)10(12)14-2/h3-6H,1-2H3

InChI key

NIQCNGHVCWTJSM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Dimethyl phthalate in mainly used in food packaging and as a solvent for fragrance.

Disclaimer

For R&D or non-EU Food use. Not for retail sale.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

294.8 °F - closed cup

Flash Point(C)

146.0 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Analysis of phthalates in food products and packaging materials sold on the Belgian market.
Fierens T, et al.
Food And Chemical Toxicology, 50(7), 2575-2583 (2012)
Food packaging and bisphenol A and bis (2-ethyhexyl) phthalate exposure: findings from a dietary intervention.
Rudel RA, et al.
Environmental Health Perspectives, 119(7), 914-914 (2001)
Guy Bouchoux et al.
The journal of physical chemistry. A, 114(27), 7408-7416 (2010-06-22)
Aromatic substitution reactions of selected ionized benzenes derivatives (chlorobenzene, nitrobenzene, dimethylphtalates) and phenoxy cation using neutral methyl isocyanide as a nucleophile are shown to efficiently occur in the gas phase. Nitrilium ions are produced in high abundance during these processes.
Yi-Hung Chen et al.
Journal of hazardous materials, 192(3), 1017-1025 (2011-07-05)
The removal of dimethyl phthalate (DMP), which is a pollutant of concern in water environments, was carried out by catalytic ozonation with TiO(2)/Al(2)O(3) catalysts. The heterogeneous catalytic ozonation was an ozonation process combined with the catalytic and adsorptive properties of
Zhujian Huang et al.
Journal of hazardous materials, 246-247, 70-78 (2013-01-05)
The organic layered double hydroxides (LHDs)/TiO(2) composites with various mass ratios were prepared by the reconstruction of mixed metal oxides to photodegrade dimethyl phthalate (DMP). The physicochemical properties of the obtained products were analyzed by X-ray diffraction (XRD) spectra, X-ray

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service