Skip to Content
Merck
All Photos(1)

Key Documents

W220841

Sigma-Aldrich

α-Isobutylphenethyl alcohol

98%

Synonym(s):

4-Methyl-1-phenyl-2-pentanol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H18O
CAS Number:
Molecular Weight:
178.27
FEMA Number:
2208
EC Number:
Council of Europe no.:
2031c
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
2.065
NACRES:
NA.21

biological source

synthetic

Quality Level

Agency

meets purity specifications of JECFA

reg. compliance

FDA 21 CFR 172.515

Assay

98%

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

green; floral; herbaceous

SMILES string

CC(C)CC(O)Cc1ccccc1

InChI

1S/C12H18O/c1-10(2)8-12(13)9-11-6-4-3-5-7-11/h3-7,10,12-13H,8-9H2,1-2H3

InChI key

IUADYGVMSDKSMB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Taste at 10 ppm

Disclaimer

For R&D or non-EU Food use. Not for retail sale.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

>230.0 °F

Flash Point(C)

> 110 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Clement Yedjou et al.
International journal of environmental research and public health, 5(5), 337-341 (2009-01-20)
Breast cancer is the leading cause of death among women between 40 and 55 years of age and is the second overall cause of death among women. Fortunately, the mortality rate from breast cancer has decreased in recent years due
Ei Kinai et al.
Journal of medical virology, 79(8), 1113-1119 (2007-06-29)
The efficacy of interferon therapy for hepatitis C virus (HCV) infection improved remarkably. However, virologic relapse occurs in a substantial proportion of patients with virologic response (defined as an HCV RNA level below 50 IU/ml at the end-of-treatment). A highly
G P Ford et al.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 21(4), 441-447 (1983-08-01)
Groups of 15 male and 15 female rats were given diets to provide intakes of 0 (control), 10, 40 or 160 mg 4-methyl-1-phenylpentan-2-ol/kg/day for 13 wk. No effects attributable to treatment were seen in the 10-mg/kg/day dose group. At 40
J Całka et al.
Polish journal of veterinary sciences, 12(1), 61-67 (2009-05-23)
Previous immunocytochemical studies provided conflicting data concerning occurrence of the CGRP-immunoreactive neuronal cell bodies in the porcine spinal cord. In the present study, we have investigated expression of the CGRP and its possible coexpression with ChAT in the gray matter
F Delgado et al.
Veterinary microbiology, 134(3-4), 383-387 (2008-10-08)
Paratuberculosis or Johne's disease is a chronic infectious disorder caused by Mycobacterium avium subsp. paratuberculosis (Map). The disease produces diarrhea and weight loss in cattle and other animal species, and it is characterized by granulomatous enteritis and lymphadenitis. Histopathology and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service