Skip to Content
Merck
All Photos(1)

Key Documents

W208604

Sigma-Aldrich

trans-Anethole

≥99%, FCC, FG

Synonym(s):

4-Propenylanisole, trans-1-Methoxy-4-(1-propenyl)benzene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH=CHC6H4OCH3
CAS Number:
Molecular Weight:
148.20
FEMA Number:
2086
Beilstein:
774229
EC Number:
Council of Europe no.:
183
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
4.010
NACRES:
NA.21
Organoleptic:
anise; sweet
grade:
FG
Fragrance grade
Halal
Kosher
biological source:
synthetic
Agency:
follows IFRA guidelines
meets purity specifications of JECFA
food allergen:
no known allergens

biological source

synthetic

Quality Level

grade

FG
Fragrance grade
Halal
Kosher

Agency

follows IFRA guidelines
meets purity specifications of JECFA

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 178/2002
FCC
FDA 21 CFR 117
FDA 21 CFR 182.60

Assay

≥99%

form

liquid

refractive index

n20/D 1.561 (lit.)

bp

234-237 °C (lit.)

mp

20-21 °C (lit.)

density

0.988 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

trans-anethole

Organoleptic

anise; sweet

SMILES string

COc1ccc(\C=C\C)cc1

InChI

1S/C10H12O/c1-3-4-9-5-7-10(11-2)8-6-9/h3-8H,1-2H3/b4-3+

InChI key

RUVINXPYWBROJD-ONEGZZNKSA-N

Looking for similar products? Visit Product Comparison Guide

Application


  • Anti-oral streptococci and anti-biofilm properties of Etlingera pavieana essential oil and its bioactive compounds proposed for an alternative herbal mouthwash.: This study evaluates the antimicrobial properties of Etlingera pavieana essential oil, identifying trans-anethole as a key bioactive compound with potential applications in oral hygiene products (Wongsariya et al., 2024).

  • Efficacy and safety of aniseed powder for treating gastrointestinal symptoms of COVID-19: a randomized, placebo-controlled trial.: The trial highlights the therapeutic potential of aniseed powder, rich in trans-anethole, for alleviating gastrointestinal symptoms in COVID-19 patients (Mosaffa-Jahromi et al., 2024).

  • Transcriptomic and biochemical analyses revealed antifungal mechanism of trans-anethole on Aspergillus flavus growth.: This research elucidates the antifungal mechanism of trans-anethole against Aspergillus flavus, suggesting its potential application in controlling fungal contamination in food and agriculture (Lei et al., 2023).

Biochem/physiol Actions

Naturally occurring phenylpropene derivative that is estrogenic at lower concentrations and cytotoxic at higher concentrations to cancer cell lines. The cytotoxicity is related to the metabolism of trans-anethole to 4-hydroxy-1-propenylbenzene.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Skin Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

213.8 °F

Flash Point(C)

101 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rajesh K Joshi et al.
Natural product communications, 6(1), 141-143 (2011-03-04)
The essential oil of the leaves of Feronia elephantum Corr. was analyzed by gas chromatography and gas chromatography/mass spectrometry. The main constituents were beta-pinene (28.4%), Z-anethole (22.1%), methyl chavicol (12.0%) and E-anethole (8.1%), among thirty-three identified compounds, which represented 92.6%
Bülent Cetin et al.
Journal of medicinal food, 13(1), 196-204 (2010-02-09)
The objective of this study was to determine the chemical compositions of the essential oil and hexane extract isolated from the inflorescence, leaf stems, and aerial parts of Florence fennel and the antimicrobial activities of the essential oil, hexane extract
Masahiro Yutani et al.
Phytotherapy research : PTR, 25(11), 1707-1713 (2011-07-02)
trans-Anethole (anethole), a major component of anise oil, has a broad antimicrobial spectrum with antimicrobial activity relatively weaker than those of well-known antibiotics, and significantly enhances the antifungal activity of polygodial and dodecanol against the baker's yeast Saccharomyces cerevisiae and
Hong-Ik Cho et al.
Journal of natural products, 76(9), 1717-1723 (2013-08-22)
The aim of this study was to investigate the hepatoprotective effect of anethole (trans-anethole, 1), a major component of Foeniculum vulgare, and its molecular mechanism during ischemia/reperfusion (I/R). Mice were subjected to 60 min of partial hepatic ischemia followed by
Meher U Nessa et al.
Anticancer research, 32(11), 4843-4850 (2012-11-17)
Chemopreventative phytochemicals having antitumour and antioxidant properties can overcome problems of chemoresistance and nonspecific toxicity towards normal cells that are associated with platinum-based chemotherapy against cancer. These agents exert their effects by bringing into play numerous cellular proteins that in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service