SYNB300125C
Synthware™ gas washing bottle with fritted disc
125 mL, joint: ST/NS 45/50, Disc porosity C
Synonym(s):
Synthware B300125C, Drechsel bottle, gas washing bottle
All Photos(1)
About This Item
UNSPSC Code:
41103209
NACRES:
NB.51
Recommended Products
material
clear bottle
cylindrical bottle
straight bottle
Joint
joint: ST/NS45/50
feature
cap
packaging
pkg of 1 ea
manufacturer/tradename
Synthware B300125C
H
290 mm
capacity
125 mL
125 mL
Related Categories
General description
Gas washing bottle with standard taper 45/50 cap style stopper and fritted disc angled at 15° for even distribution of gas throughout absorbing material. Available with a coarse or extra coarse fritted disc. Order plastic clip separately. (not supplied)
Legal Information
Synthware is a trademark of Kemtech America Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)