Skip to Content
Merck
All Photos(3)

Key Documents

M16806

Sigma-Aldrich

2-Methoxy-4-morpholinobenzenediazonium chloride zinc chloride double salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H14N3O2 · 0.5 Cl4Zn
CAS Number:
Molecular Weight:
323.85
EC Number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

form

dry powder

Quality Level

mp

131 °C (dec.) (lit.)

storage temp.

2-8°C

SMILES string

[Cl-].[Cl-].Cl[Zn]Cl.COc1cc(ccc1[N+]#N)N2CCOCC2.COc3cc(ccc3[N+]#N)N4CCOCC4

InChI

1S/2C11H14N3O2.4ClH.Zn/c2*1-15-11-8-9(2-3-10(11)13-12)14-4-6-16-7-5-14;;;;;/h2*2-3,8H,4-7H2,1H3;4*1H;/q2*+1;;;;;+2/p-4

InChI key

UKEGNQLCNXRWNG-UHFFFAOYSA-J

Application

2-Methoxy-4-morpholinobenzenediazonium chloride zinc chloride double salt can be used as an aromatic azide precursor to synthesize 1,2,3-triazole products via one-pot tandem click reactions with terminal alkynes.
Reactant used in multi-step click reactions for preparation of azides

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service