JKA7015
4arm-PEG10K
average MN 10,000, Succinimidyl Carboxymethyl Ester, amine reactive
Synonym(s):
4arm-PEG-Succinimidyl Carboxymethyl Ester, 4arm-PEG-SCM, Polyethylene glycol
About This Item
Recommended Products
Product Name
4arm-PEG10K-Succinimidyl Carboxymethyl Ester, average Mn 10,000
form
solid
mol wt
average Mn 10,000
reaction suitability
reactivity: amine reactive
polymer architecture
shape: 4-arm
functionality: homofunctional
storage temp.
−20°C
General description
Application
Legal Information
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Articles
Immune system defends against disease by hydrolyzing foreign molecules; proteases, nucleases, liver, and kidneys are involved.
Immune system defends against disease by hydrolyzing foreign molecules; proteases, nucleases, liver, and kidneys are involved.
Immune system defends against disease by hydrolyzing foreign molecules; proteases, nucleases, liver, and kidneys are involved.
Immune system defends against disease by hydrolyzing foreign molecules; proteases, nucleases, liver, and kidneys are involved.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service