GF89418606
Praseodymium
foil, thickness 1.0 mm, size 50 × 50 mm, purity 99%
Synonym(s):
PR000250
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Empirical Formula (Hill Notation):
Pr
CAS Number:
Molecular Weight:
140.91
MDL number:
UNSPSC Code:
12141610
PubChem Substance ID:
NACRES:
NA.23
Assay
99%
form
foil
manufacturer/tradename
Goodfellow 894-186-06
resistivity
68 μΩ-cm, 20°C
size
50 × 50 mm
thickness
1.0 mm
bp
3520 °C (lit.)
mp
931 °C (lit.)
density
6.71 g/mL at 25 °C (lit.)
SMILES string
[Pr]
InChI
1S/Pr
InChI key
PUDIUYLPXJFUGB-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
For updated SDS information please visit www.goodfellow.com
Legal Information
Product of Goodfellow
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service