Skip to Content
Merck
All Photos(2)

Key Documents

GF77674719

Yttrium

wire reel, 0.2m, diameter 0.5mm, 99.9%

Synonym(s):

Yttrium, Y 005105

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Y
CAS Number:
Molecular Weight:
88.91
MDL number:
UNSPSC Code:
12141617
PubChem Substance ID:
NACRES:
NA.23
Pricing and availability is not currently available.

Assay

99.9%

form

wire

manufacturer/tradename

Goodfellow 776-747-19

resistivity

57 μΩ-cm, 20°C

L × diam.

0.2 m × 0.5 mm

bp

3338 °C (lit.)

mp

1522 °C (lit.)

density

4.469 g/mL at 25 °C (lit.)

SMILES string

[Y]

InChI

1S/Y

InChI key

VWQVUPCCIRVNHF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

For updated SDS information please visit www.goodfellow.com.

Legal Information

Product of Goodfellow

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Aki Aoyama et al.
Molecular cancer therapeutics, 13(12), 2978-2990 (2014-10-15)
Tivantinib (ARQ197) was first reported as a highly selective inhibitor of c-MET and is currently being investigated in a phase III clinical trial. However, as recently reported by us and another group, tivantinib showed cytotoxic activity independent of cellular c-MET
Yukiko Ishihara et al.
Analytical sciences : the international journal of the Japan Society for Analytical Chemistry, 31(8), 781-785 (2015-08-11)
With a view to enhance the sensitivity of analytical instruments used in the measurement of trace elements contained in a single cell, we have now equipped the previously reported micro-droplet injection system (M-DIS) with a desolvation system. This modified M-DIS
Xue Wang et al.
Journal of nanoscience and nanotechnology, 14(5), 3910-3913 (2014-04-17)
Nd3+ doped alpha-NaYF4 nanocrystals with size of about 15 nm were successfully synthesized through hydrothermal method. Nearly pure near-infrared to near-infrared (NIR-to-NIR) luminescence can be realized. Moreover, the excitation and the emission at 880 and 1060 nm, respectively, are away
G Scholz et al.
Dalton transactions (Cambridge, England : 2003), 44(30), 13522-13529 (2015-07-03)
Nanoscopic yttrium acetate fluorides Y(CH(3)COO)(3-z)F(z) and yttrium oxide fluorides YO(3-z)/(2)F(z )were prepared with tunable Y/F molar ratios via the fluorolytic sol-gel route. All samples were characterized by X-ray diffraction, elemental analysis and thermal analysis. In addition, local structures of all
Ryan Hickey et al.
International journal of radiation oncology, biology, physics, 88(5), 1025-1031 (2014-03-26)
Radiosensitizing chemotherapy improves the outcomes in comparison with radiation alone for gastrointestinal cancers. The delivery of radiation therapy with yttrium90 ((90)Y) radioembolization, in combination with the radiosensitizing chemotherapeutic agent capecitabine, provides the opportunity to enhance the effects of radiation on

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service