Skip to Content
Merck
All Photos(2)

Key Documents

GF64961163

Lead

microfoil, disks, 10mm, thinness 0.025μm, specific density 30.9μg/cm2, permanent mylar 3.5μm support, 99.99+%

Synonym(s):

Lead, PB004200

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Pb
CAS Number:
Molecular Weight:
207.20
MDL number:
UNSPSC Code:
12141723
PubChem Substance ID:
NACRES:
NA.23

Assay

99.99%

form

foil

manufacturer/tradename

Goodfellow 649-611-63

resistivity

20.65 μΩ-cm

bp

1740 °C (lit.)

mp

327.4 °C (lit.)

SMILES string

[Pb]

InChI

1S/Pb

InChI key

WABPQHHGFIMREM-UHFFFAOYSA-N

General description

For updated SDS information please visit www.goodfellow.com.

Legal Information

Product of Goodfellow

Pictograms

Health hazardEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Lact. - Repr. 1A - STOT RE 1 Oral

Target Organs

Central nervous system,Blood,Immune system,Kidney

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cognitive declines after deep brain stimulation are likely to be attributable to more than caudate penetration and lead location.
Takashi Morishita et al.
Brain : a journal of neurology, 137(Pt 5), e274-e274 (2014-02-13)
Min-Yi Huang et al.
Ecotoxicology and environmental safety, 104, 310-316 (2014-04-15)
Lead (Pb) is a common heavy metal in the natural environment, but its concentration has been increasing alongside widespread industrial and agricultural development in China. The dark-spotted frog Pelophylax (formerly Rana) nigromaculata (Anura: Ranidae) is distributed across East Asia and
Reply: Cognitive declines after deep brain stimulation are likely to be attributable to more than caudate penetration and lead location.
Karsten Witt et al.
Brain : a journal of neurology, 137(Pt 5), e275-e275 (2014-02-20)
Yi Yang et al.
Toxicology letters, 227(3), 172-178 (2014-04-09)
Lead is a toxic heavy metal with many recognized adverse health side effects. The central nervous system is the main target of lead toxicity. Although many studies on lead toxicity were conducted, the mechanism of lead toxicity remains uncertain. One
Jun Yoshinaga et al.
Environmental pollution (Barking, Essex : 1987), 189, 223-228 (2014-04-01)
The levels of 25 elements in house dust collected from 100 general Japanese residences were measured. Factor analysis was applied on the multi-element data to explore source of Pb (median concentration 49.1 mg/kg) in house dust. Six factors were extracted

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service