Key Documents
DWKWCL1000AD-3
WHEATON® CELLine flasks for adherent cell culture
polystyrene, flask capacity (1,000 mL), case of 3 ea
Synonym(s):
Adherent Cell Culture Flasks, CELLine Flasks, Cell Culture Flasks
About This Item
UNSPSC Code:
41122104
NACRES:
NB.13
material
molded polypropylene neck
polystyrene
screw top plastic cap
sterility
sterile; γ-irradiated
feature
with cap
packaging
case of 3 ea
manufacturer/tradename
WHEATON® WCL1000AD-3
flask capacity
(1,000 mL)
General description
CELLine AD Bioreactor Flask for high density adherent cell culture.
Media compartment with cell growth compartment for antibody and protein production.
Use with Hybridoma, Insect, CHO, NSO, HEK, BHK cell lines
  • Disposable and ready-to-use
  • High call density and high product concentration
  • Bulk media chamber allows for longer run times without media refreshing and reduces operation time
  • Decreases use of consumables
  • Cost-efficient, space saving, and stackable
  • No additional equipment required for operation
Features and Benefits
  • Ideal for high density cell culture, designed to enhance small scale bio-production for antibody and protein generation_x000D_
  • Available in 1000ml flask for suspension (CELLine 1000) or adherent (CELLine 1000-AD) cell culture and 350ml flask for suspension culture only_x000D_
  • Disposable and ready-to-use_x000D_
  • Multi-harvest operation ensures a cost efficient solution for antibody and protein production_x000D_
  • Ideal for use with cell lines such as Hybridoma, CHO, HEK, BHK, Sf9
Legal Information
WHEATON is a registered trademark of Wheaton Industries, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)