DWK73805-15415
Kimble® Closed Top Linerless Polypropylene Screw Thread Cap
15-415, white screw cap
All Photos(1)
About This Item
UNSPSC Code:
41100000
NACRES:
NB.17
Recommended Products
material
red polypropylene screw cap (Push fit)
white screw cap
packaging
pkg of 1 packs
manufacturer/tradename
Kimble® 73805-15415
fitting
thread
Application
Closure solution for vials and bottles
Features and Benefits
- Economical, one-piece construction
- Unique design provides exceptional sealing properties
- Autoclavable
- Designed from ASTM Specification E982
Legal Information
KIMBLE is a registered trademark of DWK Life Sciences
Choose from one of the most recent versions:
Certificates of Analysis (COA)