DWK07-1100
Wheaton® Microliter μlplate Shallow Polypropylene
All Photos(1)
About This Item
UNSPSC Code:
41106200
NACRES:
NB.15
Recommended Products
material
polypropylene plate
round bottom
round wells
packaging
pkg of 1 ea
manufacturer/tradename
WHEATON® 07-1100
capacity
350 μL
Related Categories
Application
Ideal for sample collection, storage, combinatorial chemistry and library applications
Features and Benefits
- Premium virgin polypropylene shallow & medium μLplates is your assurance you achieve accurate results, the first time.
Legal Information
WHEATON is a registered trademark of Wheaton Industries, Inc.