Skip to Content
Merck
All Photos(1)

Key Documents

D61407

Sigma-Aldrich

α,α-Dichlorodiphenylmethane

97%

Synonym(s):

Benzophenone dichloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6H5)2C(Cl)2
CAS Number:
Molecular Weight:
237.12
Beilstein:
1910601
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
form:
liquid
Assay:
97%

Quality Level

Assay

97%

form

liquid

refractive index

n20/D 1.605 (lit.)

bp

305 °C (lit.)

density

1.235 g/mL at 25 °C (lit.)

SMILES string

ClC(Cl)(c1ccccc1)c2ccccc2

InChI

1S/C13H10Cl2/c14-13(15,11-7-3-1-4-8-11)12-9-5-2-6-10-12/h1-10H

InChI key

OPTDDWCXQQYKGU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Supplementary Hazards

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fujun Ma et al.
Chemosphere, 233, 913-919 (2019-07-26)
Thermal desorption has been widely employed to treat soils contaminated with chlorinated organics. The off-gas of thermal desorption must be treated to avoid secondary pollution. In this study, the treatment of DDTs in thermal desorption off-gas by pulsed corona discharge

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service