Skip to Content
Merck
All Photos(1)

Key Documents

A26403

Sigma-Aldrich

mono-Methyl adipate

99%

Synonym(s):

Adipic acid monomethyl ester, Monomethyl adipate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HO2C(CH2)4CO2CH3
CAS Number:
Molecular Weight:
160.17
Beilstein:
1766411
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

99%

form

liquid

refractive index

n20/D 1.441 (lit.)

bp

162 °C/10 mmHg (lit.)

mp

7-9 °C (lit.)

density

1.081 g/mL at 25 °C (lit.)

SMILES string

COC(=O)CCCCC(O)=O

InChI

1S/C7H12O4/c1-11-7(10)5-3-2-4-6(8)9/h2-5H2,1H3,(H,8,9)

InChI key

UOBSVARXACCLLH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service