Skip to Content
Merck
All Photos(2)

Key Documents

935751

Sigma-Aldrich

Cobalt(II) sulfate heptahydrate

greener alternative

≥99.99% trace metals basis

Synonym(s):

Cobaltous sulfate heptahydrate

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352311
Assay:
≥99.99% trace metals basis
form:
solid

Quality Level

Assay

≥99.99% trace metals basis

form

solid

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

color

orange to very dark red

greener alternative category

SMILES string

O.[Co++].[O-]S([O-])(=O)=O

InChI

InChI=1S/Co.H2O4S.7H2O/c;1-5(2,3)4;;;;;;;/h;(H2,1,2,3,4);7*1H2

InChI key

BGORGFZEVHFAQU-UHFFFAOYSA-L

General description

We are committed to bringing you Greener Alternative Products, which belongs to one of the four categories of greener alternatives. This product has been enhanced for energy efficency. Click here for more information.

Application

Cobalt(II) sulfate heptahydrate can be used in different applications.

  • Cobalt(II) sulphate heptahydrate catalyses the formation of -acetamido ketones via a one-pot multi-component reaction coupling aromatic aldehyde, acetophenone, and acetyl chloride in acetonitrile.
  • It was used to synthesis the bimetallic magnetic nanomaterial CoPt3, which was used to study ferromagnetic phase transitions from solid solution alloy to chemical ordering phase using XAS.
  • Furthermore, heterogeneous OER catalysts (CoOx, CoPi, CoFeOx) were synthesized by using Cobalt(II) sulphate heptahydrate for water oxidation which has the importannce for solar water-splitting devices, water electrolyzers, and Li-air batteries.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1B Inhalation - Eye Irrit. 2 - Muta. 2 - Repr. 1B - Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ferromagnetic CoPt3 Nanowires: Structural Evolution from fcc to Ordered L12
Chen, Hao Ming
Journal of the American Chemical Society, 131 (2009)
Efficient Synthesis of ?-Acetamido Ketones Catalyzed by Cobalt Sulfate Heptahydrate under Ultrasound Irradiation.
Chen, Bao-Hua
ChemInform, 43 (2012)
Benchmarking Heterogeneous Electrocatalysts for the Oxygen Evolution Reaction
McCrory, C. C. L. et al.
Journal of the American Chemical Society, 135 (2013)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service