Skip to Content
Merck
All Photos(1)

Key Documents

926736

Sigma-Aldrich

MNBO

≥95%

Synonym(s):

N-Benzyl-N-(2-methylnaphthalen-1-yl)oxalamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H18N2O2
CAS Number:
Molecular Weight:
318.37
MDL number:
UNSPSC Code:
12352101
NACRES:
NA.21
Pricing and availability is not currently available.

Quality Level

Assay

≥95%

form

powder

mp

202-203 °C

storage temp.

−20°C

SMILES string

N(Cc3ccccc3)C(=O)C(=O)Nc1c2c(ccc1C)cccc2

InChI key

QOOOSPCWFVCKFK-UHFFFAOYSA-N

Application

MNBO is a ligand for the copper-catalyzed Ullman-Ma cross-coupling reaction.

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jie Gao et al.
Organic letters, 19(11), 2809-2812 (2017-05-23)
A class of N-(naphthalen-1-yl)-N'-alkyl oxalamides have been proven to be powerful ligands, making a coupling reaction of (hetero)aryl iodides with primary amines proceed at 50 °C with only 0.01 mol % of Cu2O and ligand as well as a coupling

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service