Skip to Content
Merck
All Photos(1)

Key Documents

925772

Sigma-Aldrich

N,N′-Tetra(4-biphenyl)benzidine

≥97%

Synonym(s):

TBBDAz

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C60H44N2
CAS Number:
Molecular Weight:
793.00
MDL number:
UNSPSC Code:
12352116
NACRES:
NA.21

description

PL: 425 nm (in DCM)

Quality Level

Assay

≥97%

form

powder

color

white to light yellow

Orbital energy

HOMO 5.3 eV 
LUMO 2.2 eV 

UV absorption

λ: 357 nm Amax: in DCM

InChI

1S/C60H44N2/c1-5-13-45(14-6-1)49-21-33-55(34-22-49)61(56-35-23-50(24-36-56)46-15-7-2-8-16-46)59-41-29-53(30-42-59)54-31-43-60(44-32-54)62(57-37-25-51(26-38-57)47-17-9-3-10-18-47)58-39-27-52(28-40-58)48-19-11-4-12-20-48/h1-44H

InChI key

WXAIEIRYBSKHDP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Having N,N′-bis-(3-methylphenyl)-N,N′-bis-phenyl-benzidine (TPD) like unit, TBBDA (BPBPA) is electron-rich. As a result, it is used as a hole-transport material in organic light-emitting diodes (OLED).
Example of OLED Device Performance:
ITO/DNTPD (60 nm)/BPBPA (30 nm)/mCBP (10 nm)/mCBP:Ir(dbi)3 (30 nm)/LG201 (35 nm)/
LiF (1 nm)/Al (200 nm)
Color: blue
EQE: Not known
Turn-On Voltage: 2.8V
Max. Luminance: ~ 9900 Cd/m2

Besides, for organic solar cell (OSCs) applications, its highest occupied molecular orbital energy value at 5.3 eV is well matched with a donor layer and its high mobility of 1.74 × 10-2 cm2/Vs brings out an easy and rapid extraction of holes from the donor material.
N,N′-Tetra(4-biphenyl)benzidine can be used as an emissive layer or hole transporting material in studies such as organic light-emitting diodes (OLEDs), photovoltaics, organic field-effect transistors (OFETs).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Superb lifetime of blue organic light-emitting diodes through engineering interface carrier blocking layers and adjusting electron leakage and an unusual efficiency variation at low electric field.
Kim T,et al.
Journal of Material Chemistry C, 6, 8472-8478 (2018)
Four times lifetime improvement of blue phosphorescent organic light-emitting diodes by managing recombination zone.
Jeon S K, et al.
Organic Electronics, 27, 202-206 (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service