Skip to Content
Merck
All Photos(2)

Key Documents

914436

Sigma-Aldrich

Fmoc-D-Lys(Biotin)-OH

≥95%

Synonym(s):

Nα-(9-Fluorenylmethyloxycarbonyl)-Nε-biotinyl-D-lysine, N2-(((9H-Fluoren-9-yl)methoxy)carbonyl)-N6-(5-((3aS,4S,6aR)-2-oxohexahydro-1H-thieno[3,4-d]imidazol-4-yl)pentanoyl)-D-lysine, Biotinylated Fmoc-protected D-lysine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C31H38N4O6S
CAS Number:
Molecular Weight:
594.72
MDL number:
UNSPSC Code:
12352209
NACRES:
NA.22

Quality Level

Assay

≥95%

form

powder

mp

184-190 °C

storage temp.

2-8°C

InChI

1S/C31H38N4O6S/c36-27(15-6-5-14-26-28-25(18-42-26)33-30(39)35-28)32-16-8-7-13-24(29(37)38)34-31(40)41-17-23-21-11-3-1-9-19(21)20-10-2-4-12-22(20)23/h1-4,9-12,23-26,28H,5-8,13-18H2,(H,32,36)(H,34,40)(H,37,38)(H2,33,35,39)/t24-,25+,26+,28+/m1/s1

InChI key

OFIBQNGDYNGUEZ-KAHNYGAISA-N

Application

Fmoc-D-Lys(Biotin)-OH is a modified lysine derivative for the preparation of biotin-labeled peptides by Fmoc SPPS. Fmoc-L-Lys(Biotin)-OH is available as cat# 8.52097.

Automate your Biotin tagging with Synple Automated Synthesis Platform (SYNPLE-SC002)

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dongyao Wang et al.
Chemical communications (Cambridge, England), 53(36), 5020-5023 (2017-04-22)
Matrine is a plant alkaloid and a major active component in the Chinese medical herb Sophora flavescens. Matrine has shown potent anti-cancer activities but its molecular target(s) and mechanism are still unknown. Using the photo-affinity labeling approach, for the first
Probing adenosine receptors using biotinylated purine derivatives.
K A Jacobson
Methods in enzymology, 184, 668-671 (1990-01-01)
Karin A Stephenson et al.
Bioconjugate chemistry, 19(5), 1087-1094 (2008-04-15)
Through the development and application of a unique approach for producing Re-metallopeptides, a new class of peptide-derived probes that are designed to target beta-amyloid plaques was developed. Derivatives of a class of beta-breaker peptides having the core sequence lvffa or
K A Jacobson et al.
Biochemical pharmacology, 36(10), 1697-1707 (1987-05-15)
Derivatives of adenosine receptor agonists (N6-phenyladenosines) and antagonists (1,3-dialkyl-8-phenylxanthines) bearing functionalized chains suitable for attachment to other molecules have been reported [Jacobson et al., J. med. Chem. 28, 1334 and 1341 (1985)]. The "functionalized congener" approach has been extended to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service