Skip to Content
Merck
All Photos(1)

Key Documents

914096

Sigma-Aldrich

1,3-Bis(2,6-di(3-pentyl)phenyl)imidazolium chloride

≥95%

Synonym(s):

IPent-HCl

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C35H53ClN2
CAS Number:
Molecular Weight:
537.26
MDL number:
UNSPSC Code:
12352005
NACRES:
NA.22

Quality Level

Assay

≥95%

form

powder

mp

362-364 °C

SMILES string

[Cl-].[n+]2(c[n](cc2)c3c(cccc3C(CC)CC)C(CC)CC)c1c(cccc1C(CC)CC)C(CC)CC

InChI

1S/C35H53N2.ClH/c1-9-26(10-2)30-19-17-20-31(27(11-3)12-4)34(30)36-23-24-37(25-36)35-32(28(13-5)14-6)21-18-22-33(35)29(15-7)16-8;/h17-29H,9-16H2,1-8H3;1H/q+1;/p-1

InChI key

ODQOSTRWDUNLQB-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

1,3-Bis(2,6-di(3-pentyl)phenyl)imidazolium chloride is an NHC ligand used in a variety of Pd catalyzed cross-coupling reactions.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service