Skip to Content
Merck
All Photos(3)

Key Documents

902144

Sigma-Aldrich

[(Cp)Ru(η6-naphthalene)]OTf

≥95%

Synonym(s):

5-Cyclopentadienyl)(η6-naphthalene)ruthenium(1+) trifluoromethanesulfonate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H13F3O3RuS
CAS Number:
Molecular Weight:
443.40
MDL number:
UNSPSC Code:
12352101

Assay

≥95%

form

powder

reaction suitability

reagent type: catalyst
reaction type: C-C Bond Formation

mp

112-116 °C

storage temp.

2-8°C

related product

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jens Rickmeier et al.
Angewandte Chemie (International ed. in English), 57(43), 14207-14211 (2018-09-07)
Radiolabeled receptor-binding peptides are an important class of positron emission tomography tracers owing to achievable high binding affinities and their rapid blood clearance. Herein, a method to introduce a 4-[18 F]fluoro-phenylalanine residue into peptide sequences is reported, by chemoselective radio-deoxyfluorination

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service