Skip to Content
Merck
All Photos(1)

Key Documents

900517

Sigma-Aldrich

B-[4-(1,2,2-Triphenylethenyl)phenyl]boronic acid

Synonym(s):

1,2,2-Triphenylethenyl-(4′-phenylene) boronic acid, [4-(1,2,2-Triphenylvinyl)phenyl]boronic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H21BO2
CAS Number:
Molecular Weight:
376.25
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.23

Assay

97%

Quality Level

form

powder

mp

126-131 °C

SMILES string

B(O)(O)c1ccc(cc1)C(=C(c4ccccc4)c3ccccc3)c2ccccc2

InChI

1S/C26H21BO2/c28-27(29)24-18-16-23(17-19-24)26(22-14-8-3-9-15-22)25(20-10-4-1-5-11-20)21-12-6-2-7-13-21/h1-19,28-29H

InChI key

XSIVQWOJIOHPIZ-UHFFFAOYSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jiayun Xiang et al.
ACS applied materials & interfaces, 7(27), 14965-14974 (2015-06-23)
Fluorescent organic dots are emerging as promising bioimaging reagents because of their high brightness, good photostability, excellent biocompatibility, and facile surface functionalization. Organic dots with large two-photon absorption (TPA) cross sections are highly desired for two-photon fluorescence microscopy. In this
Effect of the cyano group on solid state photophysical behavior of tetraphenylethene substituted benzothiadiazoles.
Jadhav T, et al.
Journal of Material Chemistry C, 3(35), 9063-9068 (2015)
Cesar F Azael Gomez-Duran et al.
ACS applied materials & interfaces, 7(28), 15168-15176 (2015-06-26)
A series of BODIPY derivatives with tetraphenylethene (TPE) moieties were designed and synthesized. The effect of positions and numbers of substitution groups on the fluorescence of the BODIPYs was investigated. Theoretical calculation and single crystal structures proved that the TPE

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service