Skip to Content
Merck
All Photos(2)

Key Documents

805459

Sigma-Aldrich

DPEPO

Synonym(s):

Bis[2-(diphenylphosphino)phenyl] ether oxide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C36H28O3P2
CAS Number:
Molecular Weight:
570.55
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Assay

98%

Quality Level

form

powder

mp

276-281 °C

Orbital energy

HOMO 6.1 eV 
LUMO 2.0 eV 

OLED Device Performance

ITO/a-NPD/mCP/ bis-PXZ-OXD (6 wt%):DPEPO/DPEPO/TPBi/LiF/Al

  • Max. EQE: 14.9 %

ITO/a-NPD/mCP/ bis-PXZ-TDZ (6 wt%):DPEPO/DPEPO/TPBi/LiF/Al
  • Max. EQE: 10.0 %

SMILES string

O=P(C1=CC=CC=C1)(C2=CC=CC=C2)C3=C(OC4=CC=CC=C4P(C5=CC=CC=C5)(C6=CC=CC=C6)=O)C=CC=C3

InChI

1S/C36H28O3P2/c37-40(29-17-5-1-6-18-29,30-19-7-2-8-20-30)35-27-15-13-25-33(35)39-34-26-14-16-28-36(34)41(38,31-21-9-3-10-22-31)32-23-11-4-12-24-32/h1-28H

InChI key

ATTVYRDSOVWELU-UHFFFAOYSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kuan-Yu Liao et al.
Inorganic chemistry, 54(8), 4029-4038 (2015-04-08)
Tetradentate bis(pyridyl azolate) chelates are assembled by connecting two bidentate 3-trifluoromethyl-5-(2-pyridyl)azoles at the six position of pyridyl fragment with the tailored spiro-arranged fluorene and/or acridine functionalities. These new chelates were then utilized in synthesizing a series of Pt(II) metal complexes
Effect of Atom Substitution in Chalcogenodiazole-Containing Thermally Activated Delayed Fluorescence Emitters on Radiationless Transition
Tanaka et al.
The Journal of Physical Chemistry C, 119(6), 2948-2955 (2015)
Efficient blue organic light-emitting diodes employing thermally activated delayed fluorescence
Zhang et al.
Nature Photonics, 8(4), 326-332 (2014)
Qisheng Zhang et al.
Journal of the American Chemical Society, 134(36), 14706-14709 (2012-08-31)
Efficient thermally activated delayed fluorescence (TADF) has been characterized for a carbazole/sulfone derivative in both solutions and doped films. A pure blue organic light emitting diode (OLED) based on this compound demonstrates a very high external quantum efficiency (EQE) of
Mounggon Kim et al.
Advanced materials (Deerfield Beach, Fla.), 27(15), 2515-2520 (2015-03-11)
High quantum efficiency above 18% and extended lifetime three times longer than that of phosphorescent organic light-emitting diodes (OLEDs) are demonstrated in blue thermally activated delayed fluorescent OLEDs.

Articles

Organic Light-emitting Diodes (OLEDs) are solid-state devices that transform electrical energy into light. OLEDs are considered the next generation technology for high-resolution flexible displays and solid state lighting, attracting intense scientific and industrial interest.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service