Skip to Content
Merck
All Photos(1)

Key Documents

764701

Sigma-Aldrich

2,5-Dioxo-1-pyrrolidinyl 5-[4-(1,2,4,5-tetrazin-3-yl)benzylamino]-5-oxopentanoate

≥90%

Synonym(s):

5-Oxo-5-[[[4-(1,2,4,5-tetrazin-3-yl)phenyl]methyl]amino]pentanoic acid 2,5-dioxo-1-pyrrolidinyl ester, Benzylamino tetrazine N-hydroxysuccinimidyl ester, Benzylaminotetrazine-NHS, Bz-Tz-NHS

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H18N6O5
CAS Number:
Molecular Weight:
398.37
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.22

Assay

≥90%

form

solid

reaction suitability

reaction type: click chemistry
reagent type: linker

functional group

NHS ester

storage temp.

−20°C

SMILES string

O=C(CCCC(ON1C(CCC1=O)=O)=O)NCC(C=C2)=CC=C2C3=NN=CN=N3

InChI

1S/C18H18N6O5/c25-14(2-1-3-17(28)29-24-15(26)8-9-16(24)27)19-10-12-4-6-13(7-5-12)18-22-20-11-21-23-18/h4-7,11H,1-3,8-10H2,(H,19,25)

InChI key

QDUUJWMNSUXICN-UHFFFAOYSA-N

Application

2,5-Dioxo-1-pyrrolidinyl 5-[4-(1,2,4,5-tetrazin-3-yl)benzylamino]-5-oxopentanoate may be used in the synthesis of Tz-PEG11-1,4,7-triazacyclononane-N,N,N-triacetate, a precursor for Al[18F]-NOTA-labeled tetrazine radio ligand useful in 18F-based pretargeted PET imaging system.
Succinimidyl ester functionalized tetrazine for inverse electron demand Diels-Alder cycloaddition reactions. Succinimidyl ester will react with amines for small molecule, biomolecule or surface modification. Tetrazine will react with strained alkenes such as transcyclooctene, norbornene and cyclopropene to yield a stable covalent linkage. Tetrazines have proven useful in bioorthogonal reactions for many biological imaging and bioconjugation applications.

related product

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rosemery Membreno et al.
Journal of visualized experiments : JoVE, (143) (2019-02-19)
While radioimmunotherapy (RIT) is a promising approach for the treatment of cancer, the long pharmacokinetic half-life of radiolabeled antibodies can result in high radiation doses to healthy tissues. Perhaps not surprisingly, several different strategies have been developed to circumvent this
Specific Binding of Liposomal Nanoparticles through Inverse Electron?Demand Diels?Alder Click Chemistry.
Brand C, et al.
ChemistryOpen null
18F-Based Pretargeted PET Imaging Based on Bioorthogonal Diels?Alder Click Chemistry
Meyer JP, et al.
Bioconjugate Chemistry, 27(2), 298-301 (2015)
18F-Labeling of Mannan for Inflammation Research with Positron Emission Tomography.
Li XG, et al.
ACS Medicinal Chemistry Letters, 7(9), 826-830 (2016)
Mark R Karver et al.
Bioconjugate chemistry, 22(11), 2263-2270 (2011-09-29)
1,2,4,5-Tetrazines have been established as effective dienes for inverse electron demand [4 + 2] Diels-Alder cycloaddition reactions with strained alkenes for over 50 years. Recently, this reaction pair combination has been applied to bioorthogonal labeling and cell detection applications; however

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service