707230
tert-Butyldichlorophosphine solution
1.0 M in diethyl ether
Synonym(s):
(1,1-Dimethylethyl)phosphonous dichloride solution
About This Item
Recommended Products
form
liquid
reaction suitability
reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: ligand
concentration
1.0 M in diethyl ether
density
0.766 g/mL at 25 °C
functional group
phosphine
SMILES string
CC(C)(C)P(Cl)Cl
InChI
1S/C4H9Cl2P/c1-4(2,3)7(5)6/h1-3H3
InChI key
NMJASRUOIRRDSX-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Application
- The preparation of Buchwald-type secondary phosphine oxide ligands
- Asymmetric hydrogenation
Reactant for the synthesis of:
- Alkali metal complexes
- P-Stereogenic compounds via kinetic deprotonation
- Ortho-trifluoromethylphenyl-substituted phosphines
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 1 - Skin Corr. 1B - STOT SE 3
Target Organs
Respiratory system
Supplementary Hazards
Storage Class Code
3 - Flammable liquids
WGK
WGK 3
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service