Skip to Content
Merck
All Photos(1)

Key Documents

658103

Sigma-Aldrich

5-Thiazolecarboxaldehyde

95%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H3NOS
CAS Number:
Molecular Weight:
113.14
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

form

solid

refractive index

n20/D 1.5874

bp

92-94 °C/16 mmHg

density

1.304 g/mL at 25 °C

SMILES string

O=Cc1cncs1

InChI

1S/C4H3NOS/c6-2-4-1-5-3-7-4/h1-3H

InChI key

ZXRLWHGLEJGMNO-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

208.4 °F - closed cup

Flash Point(C)

98 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Raju Khatri et al.
Journal of pharmaceutical sciences, 110(1), 412-421 (2020-09-16)
Pregnancy-related hormones (PRH) have emerged as key regulators of hepatic cytochrome P450 (CYP) enzyme expression and function. The impact of PRH on protein levels of CYP3A4 and other key CYP enzymes, and the metabolism of nifedipine (a CYP3A4 substrate commonly
Asmita Pant et al.
Toxicology and applied pharmacology, 365, 61-70 (2019-01-07)
Non-alcoholic fatty liver disease is manifested by hepatic accumulation of triglycerides (TG) and is commonly associated with metabolic syndrome. The isoprenoid farnesol (FOH) modulates lipid metabolism and reduces hepatic TG content in rodents. This effect involves activation of at least
Yiqiang Xie et al.
Pakistan journal of pharmaceutical sciences, 31(5(Special)), 2315-2321 (2018-11-23)
Feng-Liao-Chang-Wei-Kang (FLCWK), a traditional Chinese patent medicine, consists primarily of Polygonum hydropiper and Daphniphyllum calycinum roots. As a complex containing several kinds of flavonoids, FLCWK has the potential to impact the drug metabolism enzyme P450 3A4 (CYP3A4) and nuclear receptors.

Articles

Synthesis of selective aldosterone synthase inhibitors using Wittig reaction with heterocyclic aldehydes described in study by Hartmann et al.

Synthesis of selective aldosterone synthase inhibitors using Wittig reaction with heterocyclic aldehydes described in study by Hartmann et al.

Synthesis of selective aldosterone synthase inhibitors using Wittig reaction with heterocyclic aldehydes described in study by Hartmann et al.

Synthesis of selective aldosterone synthase inhibitors using Wittig reaction with heterocyclic aldehydes described in study by Hartmann et al.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service