654329
6-(1-Piperazinyl)pyridine-3-boronic acid pinacol ester
97%
Synonym(s):
1-(5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-2-yl)piperazine
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C15H24BN3O2
CAS Number:
Molecular Weight:
289.18
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Quality Level
Assay
97%
form
solid
mp
137-142 °C
SMILES string
CC1(C)OB(OC1(C)C)c2ccc(nc2)N3CCNCC3
InChI
1S/C15H24BN3O2/c1-14(2)15(3,4)21-16(20-14)12-5-6-13(18-11-12)19-9-7-17-8-10-19/h5-6,11,17H,7-10H2,1-4H3
InChI key
MEHCIETYEUUYGC-UHFFFAOYSA-N
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service