Skip to Content
Merck
All Photos(1)

Key Documents

560146

Sigma-Aldrich

4-Chloro-2-iodobenzoic acid

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H3Cl(I)COOH
CAS Number:
Molecular Weight:
282.46
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

mp

163-167 °C (lit.)

functional group

carboxylic acid
chloro
iodo

SMILES string

OC(=O)c1ccc(Cl)cc1I

InChI

1S/C7H4ClIO2/c8-4-1-2-5(7(10)11)6(9)3-4/h1-3H,(H,10,11)

InChI key

LRRDANNSUCQNDU-UHFFFAOYSA-N

General description

4-Chloro-2-iodobenzoic acid is an electron-deficient substituted 2-iodobenzoic acid. It reacts with ynamide to provide the 3,4-disubstituted isocoumarin.

Application

4-Chloro-2-iodobenzoic acid may be used to synthesize:
  • 6-chloro-3-(4-methoxyphenyl)-1H-isochromen-1-one
  • (Z)-3-benzylidene-5-chloroisobenzofuran-1(3H)-one
  • 6-chloro-3-pentyl-1H-isochromen-1-one
  • 4-chloro-2-iodobenzophenone

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Regioselective synthesis of 3, 4-disubstituted isocoumarins through the Pd-catalyzed annulation of 2-iodoaromatic acids with ynamides.
Liu H, et al.
Chemical Communications (Cambridge, England), 52(41), 6801-6804 (2016)
The synthesis and electronic absorption spectra of 3-phenyl-3 (4-pyrrolidino-2-substituted phenyl)-3H-naphtho [2, 1-b] pyrans: further exploration of the ortho substituent effect.
Gabbutt CD, et al.
Tetrahedron, 62(4), 737-745 (2006)
Veronika Hampl et al.
Scientia pharmaceutica, 79(1), 21-30 (2011-05-28)
New isocoumarins were prepared in an efficient way from 2-iodobenzoic acid derivatives and hept-1-yne in a Sonogashira reaction, followed by spontaneous cyclization. Catalytic hydrogenation gave the corresponding dihydroisocoumarins. A 4-chloroisocoumarin was prepared on an alternative pathway. Some of the new
Regioselective One-Pot Synthesis of Isocoumarins and Phthalides from 2-Iodobenzoic Acids and Alkynes by Temperature Control.
Kumar MR, et al.
Advanced Synthesis & Catalysis, 355(16), 3221-3230 (2013)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service