Skip to Content
Merck
All Photos(1)

Key Documents

559091

Sigma-Aldrich

Ethyl (2-chlorobenzoyl)acetate

≥95%

Synonym(s):

Ethyl (o-chlorobenzoyl)acetate, Ethyl 3-(2-chlorophenyl)-3-oxopropanoate, NSC 158136

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClC6H4COCH2CO2C2H5
CAS Number:
Molecular Weight:
226.66
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Assay:
≥95%

Quality Level

Assay

≥95%

refractive index

n20/D 1.540 (lit.)

bp

221-222 °C (lit.)

density

1.206 g/mL at 25 °C (lit.)

functional group

chloro
ester
ketone

SMILES string

CCOC(=O)CC(=O)c1ccccc1Cl

InChI

1S/C11H11ClO3/c1-2-15-11(14)7-10(13)8-5-3-4-6-9(8)12/h3-6H,2,7H2,1H3

InChI key

DLFBNTUSDQSFOF-UHFFFAOYSA-N

Application

Ethyl (2-chlorobenzoyl)acetate may be used to synthesize ketosplitomicin derivative and ethyl 6-(2-chlorophenyl)-2,2-dimethyl-4-oxo-2,3-dihydro-4Hpyran-5-carboxylate.
Reactant for:
  • Cerium ammonium nitrate-mediated oxidative coupling
  • Hydrosilylation reactions
  • Preparation of diaryl-substituted pyrazoles as potent CCR2 receptor antagonists
  • Preparation of potential herbicidal agents
  • Ruthenium-catalyzed asymmetric hydrogenation

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Simple access to 5-carboalkoxy-2, 3-dihydro-4H-pyran-4-ones via domino acylative electrocyclization: the first three step total synthesis of the dihydronaphthopyran-4-one class of natural products.
Ilangovan A and Sakthivel P.
Royal Society of Chemistry Advances, 4(98), 55150-55161 (2014)
Characterization of sirtuin inhibitors in nematodes expressing a muscular dystrophy protein reveals muscle cell and behavioral protection by specific sirtinol analogues.
Pasco MY, et al.
Journal of Medicinal Chemistry, 53(3), 1407-1411 (2009)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service