Skip to Content
Merck
All Photos(1)

Key Documents

52435

Sigma-Aldrich

cis-1,4-Hexadiene

≥99.0%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H10
CAS Number:
Molecular Weight:
82.14
Beilstein:
1839519
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥99.0%

refractive index

n20/D 1.415

bp

65-66 °C (lit.)

density

0.707 g/mL at 20 °C (lit.)

functional group

allyl

SMILES string

C\C=C/CC=C

InChI

1S/C6H10/c1-3-5-6-4-2/h3-4,6H,1,5H2,2H3/b6-4-

InChI key

PRBHEGAFLDMLAL-XQRVVYSFSA-N

Related Categories

General description

cis-1,4-Hexadiene can be synthesized from the cobalt-catalyzed reaction between butadiene and ethylene. The reaction of this hydrocarbon with Fe(CO)5 affords the trans-1,3-hexadiene-iron tricarbonyl complex (I), plus a small amount of the trans,trans-2,4-hexadiene complex(II). Mechanism of the conversion of cis-1,4-hexadiene to trans-2-methyl-1,3-pentadiene, via rearrangement in the presence of a nickel catalyst has been investigated.

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 3 Oral - Asp. Tox. 1 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

-50.8 °F - closed cup

Flash Point(C)

-46 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nickel-promoted methylvinylcyclopropane rearrangements. Mechanistic relevance to the cis-1, 4-hexadiene to 2-methyl-1, 3-pentadiene isomerization.
Pinke PA, et al.
Journal of the American Chemical Society, 96(13), 4229-4234 (1974)
Reaction of cis-1, 4-Hexadiene with Iron Pentacarbonyl.
Kuji J and Iwamoto M.
Bulletin of the Chemical Society of Japan, 41(6), 1483-1484 (1968)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service