Skip to Content
Merck
All Photos(2)

Key Documents

520721

Sigma-Aldrich

Iridium(III) chloride

99.8% trace metals basis

Synonym(s):

Iridium trichloride, Trichloroiridium

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
IrCl3
CAS Number:
Molecular Weight:
298.58
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
Assay:
99.8% trace metals basis
form:
powder or crystals
solubility:
H2O: insoluble(lit.)
aliphatic hydrocarbons: insoluble(lit.)
aqueous acid: insoluble(lit.)

Assay

99.8% trace metals basis

form

powder or crystals

impurities

≤2500.0 ppm Trace Metal Analysis

mp

763 °C (lit.)

solubility

H2O: insoluble(lit.)
aliphatic hydrocarbons: insoluble(lit.)
aqueous acid: insoluble(lit.)

SMILES string

Cl[Ir](Cl)Cl

InChI

1S/3ClH.Ir/h3*1H;/q;;;+3/p-3

InChI key

DANYXEHCMQHDNX-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Iridium(III) chloride is a crystalline inorganic solid with excellent electrical conductivity and good transmission in the infrared range. It is used in the field of catalysis and nanomaterial synthesis.

Application

Iridium(III) chloride can be used:
  • As a precursor to synthesize Iridium nanoparticles.
  • As a starting material synthesize Iridium complexes that can be used to fabricate OLEDs.
  • As a catalyst for the ring opening of epoxidesby aryl, heterocyclic, or aliphatic amines.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Effective hydrogenation of Haloaromatic nitro compounds catalysed by iridium nanoparticles deposited on multiwall carbon nanotubes
Hua-Bin Li, et al.
Synthesis and Reactivity in Inorganic, Metal-Organic, and Nano-Metal Chemistry, 46, 1499-1505 (2016)
Ditopic bis-terdentate cyclometallating ligands and their highly luminescent dinuclear iridium (III) complexes
Pierre-Henri Lanoe, et al.
Chemical Communications (Cambridge, England), 50, 6831-6834 (2014)
Aminolysis of epoxides using iridium trichloride as an efficient catalyst
Jyoti Agarwal, et al.},
Synthesis, 2009, 2790-2796 (2009)
Rajendra Prasad Paitandi et al.
European journal of medicinal chemistry, 84, 17-29 (2014-07-12)
Efficacy of the ferrocene appended piano-stool dipyrrinato complexes [(η(6)-C6H6)RuCl(fcdpm)] (1), [(η(6)-C10H14)RuCl(fcdpm)] (2), [(η(6)-C12H18)RuCl(fcdpm)] (3) [(η(5)-C5Me5)RhCl(fcdpm)] (4) and [(η(5)-C5Me5)IrCl(fcdpm)] (5) [fcdpm = 5-ferrocenyldipyrromethene] toward anticancer activity have been described. Binding of the complexes with calf thymus DNA (CT-DNA) and BSA (bovine

Articles

Plasmonic nanoparticles have unique optical properties that can be tailored to suit a variety of applications in the biotechnology1–8 and electronics9–16 industries.

Plasmonic nanoparticles have unique optical properties that can be tailored to suit a variety of applications in the biotechnology1–8 and electronics9–16 industries.

Plasmonic nanoparticles have unique optical properties that can be tailored to suit a variety of applications in the biotechnology1–8 and electronics9–16 industries.

Plasmonic nanoparticles have unique optical properties that can be tailored to suit a variety of applications in the biotechnology1–8 and electronics9–16 industries.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service