Skip to Content
Merck
All Photos(1)

Key Documents

490695

Sigma-Aldrich

Pyrene-d10

98 atom % D

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16D10
CAS Number:
Molecular Weight:
212.31
MDL number:
UNSPSC Code:
12142201
PubChem Substance ID:
NACRES:
NA.12
Pricing and availability is not currently available.

isotopic purity

98 atom % D

Quality Level

mp

145-148 °C (lit.)

format

neat

mass shift

M+10

SMILES string

[2H]c1c([2H])c2c([2H])c([2H])c3c([2H])c([2H])c([2H])c4c([2H])c([2H])c(c1[2H])c2c34

InChI

1S/C16H10/c1-3-11-7-9-13-5-2-6-14-10-8-12(4-1)15(11)16(13)14/h1-10H/i1D,2D,3D,4D,5D,6D,7D,8D,9D,10D

InChI key

BBEAQIROQSPTKN-LHNTUAQVSA-N

Looking for similar products? Visit Product Comparison Guide

Application

This product may be used as an analytical standard.

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lu Yang et al.
Journal of environmental sciences (China), 99, 72-79 (2020-11-14)
This study encompassed the regular observation of nine polycyclic aromatic hydrocarbons (PAHs) and three nitro-PAHs (NPAHs) in particulate matter (PM) in Shanghai in summer and winter from 2010 to 2018. The results showed that the mean concentrations of ƩPAHs in
Dennis S Mkhize et al.
Water science and technology : a journal of the International Association on Water Pollution Research, 79(5), 808-819 (2019-04-27)
A passive sampling device, based on molecularly imprinted membranes (MIM), was fabricated and optimised for sampling polychlorinated biphenyls (PCBs) in aquatic ecosystems. The newly-developed passive sampler was subjected to in-situ calibration studies to determine PCB sampling rates under various conditions
Inyang O Oyo-Ita et al.
Archives of environmental contamination and toxicology, 70(2), 372-382 (2015-11-08)
The distribution of aliphatic and aromatic hydrocarbons in surface sediments of the lower course of the Imo River (Nigeria) was investigated to determine the sources and fate of these compounds. The aliphatic fraction is characterized by a widespread contribution of
Wan-Ting Chang et al.
Marine pollution bulletin, 97(1-2), 217-223 (2015-06-16)
This work investigates the dependence of the sampling rate (Rs) of semi-permeable membrane devices (SPMDs) on flow rate and temperature. The in situ Rs values were obtained using performance reference compounds (PRCs) with weighted polynomial regression and used to estimate
Noor A Aly et al.
Journal of exposure science & environmental epidemiology, 31(5), 810-822 (2021-04-26)
Hurricane Florence made landfall in North Carolina in September 2018 causing extensive flooding. Several potential point sources of hazardous substances and Superfund sites sustained water damage and contaminants may have been released into the environment. This study conducted temporal analysis

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service