489697
Adipic acid-1,6-13C2
99 atom % 13C
Synonym(s):
Hexanedioc acid-13C2
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
HO213C(CH2)413CO2H
CAS Number:
Molecular Weight:
148.13
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
Recommended Products
isotopic purity
99 atom % 13C
form
solid
bp
265 °C/100 mmHg (lit.)
mp
151-154 °C (lit.)
mass shift
M+2
SMILES string
O[13C](=O)CCCC[13C](O)=O
InChI
1S/C6H10O4/c7-5(8)3-1-2-4-6(9)10/h1-4H2,(H,7,8)(H,9,10)/i5+1,6+1
InChI key
WNLRTRBMVRJNCN-MPOCSFTDSA-N
Packaging
This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Dam. 1
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Flash Point(F)
384.8 °F - closed cup
Flash Point(C)
196.00 °C - closed cup
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
S Huclier-Markai et al.
Rapid communications in mass spectrometry : RCM, 32(16), 1331-1343 (2018-05-29)
Natural organic matter (NOM) is present in the environment and could influence the migration of heavy metals/radionuclides. The dissolved fraction of NOM (DOM) is usually quantified using total organic carbon analysis or UV-visible spectrometry. Nonetheless, analysis using pattern recognition cannot
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service