Skip to Content
Merck
All Photos(1)

Key Documents

47543

Sigma-Aldrich

N-Fmoc-1,6-hexanediamine hydrobromide

≥98.0% (AT)

Synonym(s):

9-Fluorenylmethyl N-(6-aminohexyl)carbamate hydrobromide, N-Fmoc-1,6-diaminohexane hydrobromide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H26N2O2 · HBr
CAS Number:
Molecular Weight:
419.36
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.22
form:
solid
Assay:
≥98.0% (AT)

Quality Level

Assay

≥98.0% (AT)

form

solid

reaction suitability

reagent type: cross-linking reagent

mp

~153 °C (dec.)

functional group

Fmoc
amine

storage temp.

2-8°C

SMILES string

NCCCCCCNC(OCC1C2=C(C=CC=C2)C3=C1C=CC=C3)=O.[H]Br

InChI

1S/C21H26N2O2.BrH/c22-13-7-1-2-8-14-23-21(24)25-15-20-18-11-5-3-9-16(18)17-10-4-6-12-19(17)20;/h3-6,9-12,20H,1-2,7-8,13-15,22H2,(H,23,24);1H

InChI key

QSBKKENLOBODIG-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service