465348
2-Mercaptophenol
95%
Synonym(s):
2-Hydroxythiophenol
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
HSC6H4OH
CAS Number:
Molecular Weight:
126.18
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
95%
refractive index
n20/D 1.6060 (lit.)
density
1.255 g/mL at 25 °C (lit.)
SMILES string
Oc1ccccc1S
InChI
1S/C6H6OS/c7-5-3-1-2-4-6(5)8/h1-4,7-8H
InChI key
VMKYTRPNOVFCGZ-UHFFFAOYSA-N
General description
2-Mercaptophenol (H2mp) is a 1,2-bidentate sulfur containing ligand. It exhibits versatile coordination modes in its ligation to the transition metal atoms.
Application
2-Mercaptophenol may be employed as a complexing agent for the microwave-assisted extraction (MAE) of mercury species from sediments and zoobenthos samples. It may be used for the synthesis of Vanadium (IV) complexes having mixed O, S donor ligands.
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 3 Oral - Eye Irrit. 2 - Skin Corr. 1B
Storage Class Code
6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials
WGK
WGK 3
Flash Point(F)
230.0 °F - closed cup
Flash Point(C)
110 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Paul R. Klich et al.
Inorganic chemistry, 35(2), 347-356 (1996-01-17)
Reaction of VO(acac)(2) with 2-mercaptophenol (mpH(2)) in the presence of triethylamine gives the mononuclear tris complex (Et(3)NH)(2)[V(mp)(3)] (1), in which the vanadyl oxygen has been displaced. An analogous reaction using 2-mercapto-4-methylphenol (mmpH(2)) afforded (Et(3)NH)(PNP)[V(mmp)(3)] (2), which was structurally characterized. 2
Benzodiazepine Analogues. Part 15.1 Synthesis of Benzoxathiepine Derivatives.
Gelebe AC and Kaye PT.
Synthetic Communications, 29, 4459-4475 (1996)
Jana Margetínová et al.
Analytica chimica acta, 615(2), 115-123 (2008-04-30)
A high-pressure microwave digestion was applied for microwave-assisted extraction (MAE) of mercury species from sediments and zoobenthos samples. A mixture containing 3molL(-1) HCl, 50% aqueous methanol and 0.2molL(-1) citric acid (for masking co-extracted Fe(3+)) was selected as the most suitable
Palladium complexes with simultaneous O: S coordination, syntheses, structures and characterization of complexes with 2-mercaptophenol or 2-mercaptopyridine N-oxide.
Shi J-C, et al.
Polyhedron, 16(3), 369-375 (1997)
Steven Goossens et al.
Nature communications, 6, 5794-5794 (2015-01-08)
Early T-cell precursor leukaemia (ETP-ALL) is a high-risk subtype of human leukaemia that is poorly understood at the molecular level. Here we report translocations targeting the zinc finger E-box-binding transcription factor ZEB2 as a recurrent genetic lesion in immature/ETP-ALL. Using
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service