Skip to Content
Merck
All Photos(1)

Key Documents

464163

Sigma-Aldrich

1,3-Propanediol cyclic sulfate

98%

Synonym(s):

1,3,2-Dioxathiane 2,2-dioxide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C3H6O4S
CAS Number:
Molecular Weight:
138.14
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

form

solid

mp

58-62 °C (lit.)

SMILES string

O=S1(=O)OCCCO1

InChI

1S/C3H6O4S/c4-8(5)6-2-1-3-7-8/h1-3H2

InChI key

OQYOVYWFXHQYOP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Bacillus sp. CCZU11-1 catalyzed biotransformation of 1,3-propanediol cyclic sulfate (1,3-PDS) and its derivatives has been proposed. It can undergo biocatalyzed transformation to diols by Rhodococcus sp.

Application

1,3-Propanediol cyclic sulfate may be used for the synthesis of substituted (p-aminostyryl)-1-(3-sulfooxypropyl)pyridinium (ASSP) inner salts.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 2 - Eye Dam. 1 - Muta. 2 - Skin Sens. 1B

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Erratum to: Biotransformation of 1,3-propanediol cyclic sulfate and its derivatives to diols by Rhodococcus sp.
Yu-Cai He et al.
Biotechnology letters, 37(1), 189-189 (2014-10-09)
Yoshitaka Takagai et al.
Journal of chromatography. A, 1621, 461033-461033 (2020-03-20)
A diethylammonio-propylsulfate amphoteric ionic resin was synthesized and employed as the stationary phase for high-performance liquid chromatography-mass spectrometry (LC-MS) separation and detection of amino acids. The influence of experimental conditions such as mobile phase composition, column length and temperature upon
Yu-Cai He et al.
Applied biochemistry and biotechnology, 175(5), 2647-2658 (2014-12-31)
In this study, it was the first report that Bacillus sp. CCZU11-1 was used for the biotransformation of 1,3-propanediol cyclic sulfate (1,3-PDS) and its derivatives. The catalytic performance of Bacillus sp. sulfatase in the biotransformation of 1,3-PDS was significantly improved
Synthesis and properties of substituted (p-aminostyryl)-1-(3-sulfooxypropyl) pyridinium inner salts as a new class of two-photon pumped lasing dyes.
Zheng Q, et al.
Journal of Materials Chemistry, 13(10), 2499-2504 (2003)
Eric Schönemann et al.
Polymers, 11(6) (2019-06-12)
Films of zwitterionic polymers are increasingly explored for conferring fouling resistance to materials. Yet, the structural diversity of polyzwitterions is rather limited so far, and clear structure-property relationships are missing. Therefore, we synthesized a series of new polyzwitterions combining ammonium

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service