Skip to Content
Merck
All Photos(1)

Key Documents

449652

Sigma-Aldrich

Barium chloride

AnhydroBeads, −10 mesh, 99.95% trace metals basis

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
BaCl2
CAS Number:
Molecular Weight:
208.23
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

product line

AnhydroBeads

Quality Level

Assay

99.95% trace metals basis

impurities

≤550.0 ppm Trace Metal Analysis

particle size

−10 mesh

mp

963 °C (lit.)

density

3.856 g/mL at 25 °C (lit.)

SMILES string

Cl[Ba]Cl

InChI

1S/Ba.2ClH/h;2*1H/q+2;;/p-2

InChI key

WDIHJSXYQDMJHN-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Legal Information

AnhydroBeads is a trademark of Sigma-Aldrich Co. LLC

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Acute Tox. 4 Inhalation - Eye Irrit. 2

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sarah E Pierce et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 15(42), 11244-11255 (2009-09-12)
Isoguanine (2-oxo-6-amino-guanine), a natural but non-standard base, exhibits unique self-association properties compared to its isomer, guanine, and results in formation of different higher order DNA structures. In this work, the higher order structures formed by oligonucleotides containing guanine repeats or
Anne R Crecelius et al.
American journal of physiology. Heart and circulatory physiology, 307(5), H782-H791 (2014-06-29)
We tested the hypothesis that activation of inwardly rectifying potassium (KIR) channels and Na(+)-K(+)-ATPase, two pathways that lead to hyperpolarization of vascular cells, contributes to both the onset and steady-state hyperemic response to exercise. We also determined whether after inhibiting
Laura A Merriam et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 33(10), 4614-4622 (2013-03-08)
After G-protein-coupled receptor activation and signaling at the plasma membrane, the receptor complex is often rapidly internalized via endocytic vesicles for trafficking into various intracellular compartments and pathways. The formation of signaling endosomes is recognized as a mechanism that produces
Anne R Crecelius et al.
The Journal of physiology, 590(21), 5349-5359 (2012-07-11)
Circulating ATP possesses unique vasomotor properties in humans and has been hypothesized to play a role in vascular control under a variety of physiological conditions. However, the primary downstream signalling mechanisms underlying ATP-mediated vasodilatation remain unclear. The purpose of the
Michael Xavier Doss et al.
PloS one, 7(7), e40288-e40288 (2012-07-21)
Human induced pluripotent stem cell-derived cardiomyocytes (hiPSC-CM) hold promise for therapeutic applications. To serve these functions, the hiPSC-CM must recapitulate the electrophysiologic properties of native adult cardiomyocytes. This study examines the electrophysiologic characteristics of hiPSC-CM between 11 and 121 days

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service