Skip to Content
Merck
All Photos(3)

Key Documents

414336

Sigma-Aldrich

Azetidine hydrochloride

97%

Synonym(s):

Trimethyleneimine hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C3H7N · HCl
CAS Number:
Molecular Weight:
93.56
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

mp

>300 °C (lit.)

SMILES string

Cl[H].C1CNC1

InChI

1S/C3H7N.ClH/c1-2-4-3-1;/h4H,1-3H2;1H

InChI key

HGQULGDOROIPJN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

305.6 °F

Flash Point(C)

152 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Christopher S Dunkley et al.
Bioorganic & medicinal chemistry letters, 13(17), 2899-2901 (2003-11-13)
A series of compounds containing an N-(4'-substituted-3'-nitrophenyl)sydnone moiety with potential antitumor activity was prepared based on active analogues. The rationale behind the design of these compounds is presented along with the 4-step synthetic route to the derivatives in the 4'-position
Valérian Gobé et al.
Organic letters, 16(20), 5438-5441 (2014-10-01)
1,5-/1,6-Allenals conjugated to an aromatic ring undergo a cyclization, in the presence of an amine, that leads to tricyclic compounds including the 1-aminotetralin scaffold. This domino process combines the in situ formation of the enamine and the cyclization affording the
Synthesis of ornithine lactams via diastereoselective photocyclization of 2-amino-4-oxo-4-phenyl-butanoyl amines.
Lindemann U, et al.
Tetrahedron Asymmetry, 9(24), 4459-4473 (1998)
Dongliang Chang et al.
Organic letters, 4(11), 1859-1862 (2002-05-25)
[reaction: see text] Hydroxylation of N-substituted azetidines 11 and 12 and piperidines 15-19 with Sphingomonas sp. HXN-200 gave 91-98% of the corresponding 3-hydroxyazetidines 13 and 14 and 4-hydroxypiperidines 20-24, respectively, with high activity and excellent regioselectivity. High yields and high

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service