Skip to Content
Merck
All Photos(1)

Key Documents

410659

Sigma-Aldrich

4-Hydroxy-3-methoxyphenylacetone

96%

Synonym(s):

1-(4-Hydroxy-3-methoxyphenyl)-2-propanone, 2-Propiovanillone, 4-Hydroxy-3-methoxyphenyl-2-propanone, Guaiacylacetone, Methyl vanillyl ketone, Vanillyl methyl ketone

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOC6H3(OCH3)CH2COCH3
CAS Number:
Molecular Weight:
180.20
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

96%

form

liquid

refractive index

n20/D 1.55 (lit.)

bp

126-127 °C/0.3 mmHg (lit.)

density

1.163 g/mL at 25 °C (lit.)

functional group

ketone

SMILES string

COc1cc(CC(C)=O)ccc1O

InChI

1S/C10H12O3/c1-7(11)5-8-3-4-9(12)10(6-8)13-2/h3-4,6,12H,5H2,1-2H3

InChI key

LFVCJQWZGDLHSD-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Michaela Kolb et al.
Journal of chromatography. A, 1307, 144-157 (2013-08-13)
A headspace solid-phase microextraction method with subsequent GC-MS (HS-SPME/GC-MS) was established for the quantitative analysis of volatile lignin derived phenolic monomers in complex aqueous solutions. Extraction was done using a polyacrylate fiber. The optimization of HS-SPME - parameters was performed
Aqueous-phase hydrodeoxygenation of bio-derived phenols to cycloalkanes.
Zhao C, et al.
J. Catal., 280(1), 8-16 (2011)
J Jodynis-Liebert
Xenobiotica; the fate of foreign compounds in biological systems, 23(6), 693-701 (1993-06-01)
1. Metabolites of 1-(4-hydroxy-3-methoxyphenyl)-2-propanone (HMP-one), a smoke flavour compound, were isolated from rat urine using hydrolysis, ether extraction, t.l.c. and g.l.c. 2. Three metabolites were identified by mass spectrometry and independent synthesis, namely: 1-(3, 4-dihydroxyphenyl)-2-propanone (Met I), 1-(3, 4-dihydroxyphenyl)-2-propanol (Met
Mary L Forsling et al.
British journal of pharmacology, 135(3), 649-656 (2002-02-09)
Methylenedioxymethamphetamine (MDMA, 'ecstasy'), widely used as a recreational drug, can produce hyponatraemia. The possibility that this could result from stimulation of vasopressin by MDMA or one of its metabolites has been investigated in vitro. Release of both oxytocin and vasopressin
H J Helmlin et al.
Journal of analytical toxicology, 20(6), 432-440 (1996-10-01)
In Europe, the compound 3,4-methylenedioxymethamphetamine (MDMA, Ecstasy, Adam), in addition to cannabis, is the most abused illicit drug at all-night "techno" parties. Methods for the determination of MDMA and its metabolites, 4-hydroxy-3-methoxymethamphetamine (HMMA), 3,4-dihydroxy-methamphetamine (HHMA), 3,4-methylenedioxyamphetamine (MDA), 4-hydroxy-3-methoxyamphetamine (HMA), and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service