406481
Silicon phthalocyanine dihydroxide
Dye content 75 %
Synonym(s):
Dihydroxy(phthalocyaninato)silicon, Dihydroxysiliconphthalocyanine
About This Item
Recommended Products
form
solid
Quality Level
composition
Dye content, 75%
λmax
672 nm
SMILES string
O[Si]1(O)n2c3nc4nc(nc5n1c(nc6nc(nc2c7ccccc37)c8ccccc68)c9ccccc59)c%10ccccc4%10
InChI
1S/C32H18N8O2Si/c41-43(42)39-29-21-13-5-6-14-22(21)31(39)37-27-19-11-3-4-12-20(19)28(34-27)38-32-24-16-8-7-15-23(24)30(40(32)43)36-26-18-10-2-1-9-17(18)25(33-26)35-29/h1-16,41-42H/b35-25-,35-29-,36-26-,36-30-,37-27-,37-31-,38-28-,38-32-
InChI key
IKEIGECHKXPQKT-GMKHRMEJSA-N
Related Categories
Application
- Structural determination, characterization and computational studies of doped semiconductors base silicon phthalocyanine dihydroxide and dienynoic acids: This paper explores the chemical doping of silicon phthalocyanine dihydroxide and its interaction with dienynoic acids (Vergara et al., 2024).
- Facile fabrication of silicon (IV) phthalocyanine-embedded poly (vinyl alcohol)-based antibacterial and antifouling interfaces: This research focuses on the fabrication and application of silicon phthalocyanine dihydroxide in antibacterial and antifouling interfaces (Strokov et al., 2020).
- … 2-ethoxy substituted as well as axially (4-phenylpiperazin-1-yl) propanoxy-disubstituted silicon phthalocyanines and their electrochemistry: This paper details the synthesis and electrochemical properties of silicon phthalocyanine dihydroxide and its derivatives (Biyiklioglu and Alp, 2015).
- Methylsilicon phthalocyanine hydroxide doped PVA films for optoelectronic applications: This study examines the properties of methylsilicon phthalocyanine hydroxide in doped PVA films for optoelectronic uses (Qashou et al., 2019).
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service