Skip to Content
Merck
All Photos(3)

Key Documents

369144

Sigma-Aldrich

2-(Trifluoromethyl)acrylic acid

98%

Synonym(s):

TFMAA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2C=C(CF3)CO2H
CAS Number:
Molecular Weight:
140.06
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

Assay

98%

mp

51-52 °C (lit.)

SMILES string

OC(=O)C(=C)C(F)(F)F

InChI

1S/C4H3F3O2/c1-2(3(8)9)4(5,6)7/h1H2,(H,8,9)

InChI key

VLSRKCIBHNJFHA-UHFFFAOYSA-N

Application

2-(Trifluoromethyl)acrylic acid (TFMAA) is a strong acid functional monomer, which shows a good performance in the solid-phase extraction of domoic acid. It can also be used in the preparation of molecularly imprinted polymers that show diastereoselectivity for cinchona alkaloids.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

179.6 °F - closed cup

Flash Point(C)

82 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Zhengqiang Dai et al.
Journal of molecular modeling, 21(11), 290-290 (2015-10-27)
Recently, a series of computational and combinatorial approaches were employed to improve the efficiency of screening for optimal molecularly imprinted polymer (MIP) systems. In the present work, we investigated MIP systems based on enrofloxacin (ENRO) as the template molecule and
Molecularly imprinted fluorescent-shift receptors prepared with 2-(trifluoromethyl) acrylic acid
Matsui J, et al.
Analytical Chemistry, 72(14), 3286-3290 (2000)
Extraction of domoic acid from seawater and urine using a resin based on 2-(trifluoromethyl) acrylic acid
Piletska EV, et al.
Analytica Chimica Acta, 610(1), 35-43 (2008)
Porkodi Kadhirvel et al.
Analytical and bioanalytical chemistry, 411(8), 1525-1536 (2019-02-03)
A molecularly imprinted polymer (MIP) was designed in order to allow the selective solid-phase extraction of carbamazepine (CBZ), an anticonvulsant and mood-stabilizing drug, at ultra-trace level from aqueous environmental samples. A structural analog of CBZ was selected as a dummy
Ling Zhang et al.
Nature, 557(7703), 86-91 (2018-05-04)
The formation of condensed matter typically involves a trade-off between structural order and flexibility. As the extent and directionality of interactions between atomic or molecular components increase, materials generally become more ordered but less compliant, and vice versa. Nevertheless, high

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service