366560
1,4-Dibromobenzene-d4
98 atom % D
Synonym(s):
Tetradeutero-p-dibromobenzene
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Empirical Formula (Hill Notation):
C6D4Br2
CAS Number:
Molecular Weight:
239.93
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.12
Recommended Products
isotopic purity
98 atom % D
form
solid
technique(s)
protein expression: suitable
bp
219 °C
mp
88-90 °C (lit.)
density
1.872 g/mL at 25 °C
mass shift
M+4
SMILES string
[2H]c1c([2H])c(Br)c([2H])c([2H])c1Br
InChI
1S/C6H4Br2/c7-5-1-2-6(8)4-3-5/h1-4H/i1D,2D,3D,4D
InChI key
SWJPEBQEEAHIGZ-RHQRLBAQSA-N
Related Categories
Packaging
This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Aquatic Chronic 2 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 2
Flash Point(F)
212.0 °F - closed cup
Flash Point(C)
100.00 °C - closed cup
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service