Skip to Content
Merck
All Photos(3)

Documents

366315

Sigma-Aldrich

Tris(dibenzylideneacetone)dipalladium(0)-chloroform adduct

Synonym(s):

Dipalladium-tris(dibenzylideneacetone)chloroform complex, Pd2(dba)3 · CHCl3

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6H5CH=CHCOCH=CHC6H5)3Pd2 · CHCl3
CAS Number:
Molecular Weight:
1035.10
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

form

solid

Quality Level

reaction suitability

core: palladium
reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Cross Couplings
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: catalyst

mp

131-135 °C (lit.)

SMILES string

[Pd].[Pd].ClC(Cl)Cl.O=C(/C=C/c1ccccc1)\C=C\c2ccccc2.O=C(/C=C/c3ccccc3)\C=C\c4ccccc4.O=C(\C=C\c5ccccc5)/C=C/c6ccccc6

InChI

1S/3C17H14O.CHCl3.2Pd/c3*18-17(13-11-15-7-3-1-4-8-15)14-12-16-9-5-2-6-10-16;2-1(3)4;;/h3*1-14H;1H;;/b3*13-11+,14-12+;;;

InChI key

LNAMMBFJMYMQTO-FNEBRGMMSA-N

General description

Tris(dibenzylideneacetone)dipalladium(0)-chloroform adduct (Pd2dba3·CHCl3) is reported to serve as effective precatalysts, or precursors to the active Pd(0) catalyst.

Application

Tris(dibenzylideneacetone)dipalladium(0)-chloroform adduct (Pd2dba3·CHCl3) was used in the following studies:
  • To compose the catalytic system for the preparation of homoallylpalladium complexes.These complexes underwent in situ Stille type cross coupling with various vinyltin reagents to afford the cyclized products bearing allyl appendages.
  • As palladium source in the asymmetric transformations of 3,4-epoxy-1-butene.
  • As catalyst for the Heck cross-coupling reaction of iodobenzene with styrene.
  • As cyclization catalyst.
  • As catalyst for [2+2+2] cycloaddtion of didehydrotriphenylenes to the corresponding extended triphenylenes.
  • As catalyst for the carbonylation of b,b-imidoyl iodides to the corresponding imidate esters used, in turn, to prepare cyclic, quaternary amino acids.

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Carc. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tetrahedron Letters, 30, 651-651 (1989)
The Journal of Organic Chemistry, 55, 3388-3388 (1990)
Palladate salts from ionic liquids as catalysts in the Heck reaction.
Gayet M,et al.
ARKIVOC (Gainesville, FL, United States), 17, 61-76 (2008)
Palladium (II)-catalyzed tandem cyclic carbopalladation-vinylation of enyne compounds.
Yamada H, et al.
Tetrahedron Letters, 38(17), 3027-3030 (1997)
Santos Fustero et al.
Organic letters, 8(18), 4129-4132 (2006-08-25)
The synthesis of new beta,beta-difluorinated cyclic quaternary alpha-amino acid derivatives 1 in which a ring-closing metathesis reaction (RCM) constitutes the key step is described. The approach employs imidoyl chlorides 3 as fluorinated building blocks, and the overall process involves the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service