Skip to Content
Merck
All Photos(1)

Key Documents

330523

Sigma-Aldrich

(R)-(−)-2-Pentanol

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2CH2CH(OH)CH3
CAS Number:
Molecular Weight:
88.15
Beilstein:
4652311
MDL number:
UNSPSC Code:
12352001
PubChem Substance ID:
NACRES:
NA.22

vapor density

3 (vs air)

Assay

98%

form

liquid

optical activity

[α]25/D −13°, neat

autoignition temp.

657 °F

expl. lim.

9 %

refractive index

n20/D 1.406 (lit.)

bp

119-120 °C (lit.)

density

0.809 g/mL at 20 °C (lit.)

SMILES string

CCC[C@@H](C)O

InChI

1S/C5H12O/c1-3-4-5(2)6/h5-6H,3-4H2,1-2H3/t5-/m1/s1

InChI key

JYVLIDXNZAXMDK-RXMQYKEDSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Flam. Liq. 3 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

93.2 °F - closed cup

Flash Point(C)

34 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service