Recommended Products
Quality Level
Assay
97%
form
liquid
refractive index
n20/D 1.528 (lit.)
bp
141-142 °C/745 mmHg (lit.)
density
1.12 g/mL at 25 °C (lit.)
SMILES string
Cc1cncs1
InChI
1S/C4H5NS/c1-4-2-5-3-6-4/h2-3H,1H3
InChI key
RLYUNPNLXMSXAX-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
The microwave spectrum of 5-methylthiazole was investigated by employing both Fourier transform and Stark spectroscopy. 5-methyl thiazole undergoes alkylation with n-butyl bromide followed by anion exchange of Br- for BF4- to give mobile, room-temperature ionic liquids.
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
3 - Flammable liquids
WGK
WGK 3
Flash Point(F)
107.6 °F - closed cup
Flash Point(C)
42 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Thiazolium-ion based organic ionic liquids (OILs).< sup> 1, 2</sup> Novel OILs which promote the benzoin condensation.
Tetrahedron Letters, 40(9), 1621-1622 (1999)
The microwave spectrum of 5-methylthiazole: methyl internal rotation,< sup> 14</sup> N nuclear quadrupole coupling and electric dipole moment.
Journal of Molecular Structure, 190, 295-305 (1998)
Neurochemical research, 39(8), 1485-1495 (2014-05-29)
Epidemiological and experimental evidence indicated that hyperhomocysteinemia is associated with neurodegeneration. However, homocysteine neurotoxic effects have been so far investigated mostly by employing homocysteine concentrations (≥100 µM) much higher than homocysteine mean plasma levels (20 µM) observed in patients with
Tumour biology : the journal of the International Society for Oncodevelopmental Biology and Medicine, 35(8), 8301-8307 (2014-05-27)
This study was carried out to evaluate the effects of gastric cancer cell supernatant on human peritoneal mesothelial cell viability and apoptosis and to investigate the mechanism of action of gastric cancer in a mesothelial cell line (HMrSV5). Cell viability
Tumour biology : the journal of the International Society for Oncodevelopmental Biology and Medicine, 36(3), 1739-1745 (2014-11-07)
This study was carried out to evaluate the effects of a Huaier polysaccharide (TP-1) on the tumor growth and immune function in hepatocellular carcinoma (HCC) H22-based mouse in vivo. Results showed that TP-1 was capable of repressing transplanted H22 solid
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service