Skip to Content
Merck
All Photos(1)

Key Documents

295442

Sigma-Aldrich

Hydrogen sulfide

≥99.5%

Synonym(s):

Dihydrogen monosulfide, Hydrosulfuric acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
H2S
CAS Number:
Molecular Weight:
34.08
Beilstein:
3535004
EC Number:
MDL number:
UNSPSC Code:
12142100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

vapor density

1.19 (15 °C, vs air)

Quality Level

vapor pressure

252 psi ( 21 °C)

Assay

≥99.5%

autoignition temp.

500 °F

expl. lim.

6 %

bp

−60 °C (lit.)

mp

−85 °C (lit.)

SMILES string

S

InChI

1S/H2S/h1H2

InChI key

RWSOTUBLDIXVET-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Hydrogen sulfide serves as a precursor for elemental sulfur and in the production of organosulfur compounds.[1]

Application

Hydrogen sulfide gas can be used in the following applications:
  • As a sulfur source to synthesize assemblies of silver sulfide nanoparticles (Ag2S NPs).[2]
  • As a ligand to form coordination complexes with ruthenium ammines.[3]
  • As a test compound to assess the performance of polyaniline-based gas H2S sensors.[4][5]

Packaging

Supplied in a carbon steel lecture bottle with a CGA180M/CGA110F needle valve installed.

Compatible with the following:
  • Aldrich® lecture-bottle station systems
  • Aldrich® lecture-bottle gas regulators

Other Notes

Legal Information

Aldrich is a registered trademark of Sigma-Aldrich Co. LLC

hose barb

Product No.
Description
Pricing

purge valve

Product No.
Description
Pricing

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 1 Inhalation - Aquatic Acute 1 - Flam. Gas 1A - Press. Gas Liquefied gas

Storage Class Code

2A - Gases

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Printed hydrogen sulfide gas sensor on paper substrate based on polyaniline composite.
Sarfraz J
Thin Solid Films, 534, 621-628 (2013)
Fabrication of polyaniline-based gas sensors using piezoelectric inkjet and screen printing for the detection of hydrogen sulfide.
Crowley K
IEEE Sensors Journal, 10(9), 1419-1426 (2010)
Silver sulfide nanoparticle assembly obtained by reacting an assembled silver nanoparticle template with hydrogen sulfide gas.
Chen R
Nanotechnology, 19(45), 455604-455604 (2008)
Hydrogen sulfide
Pouliquen F, et al.
Ullmann's Encyclopedia of Industrial Chemistry (2000)
Amine Ruthenium complexes of hydrogen sulfide and related sulfur ligands.
Kuehn CC
Journal of the American Chemical Society, 98(3), 689-702 (1976)

Questions

1–3 of 3 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  3. Hi, I need to use it for USP ID test by bubbling H2S gas in water. Is it a liquified form? what type of container is it in? Do I need special equipment, like regulator, to use with it? Thank you

    1 answer
    1. Product 295442 is a liquified gas and is packaged in a 440 ml carbon steel lecture bottle with a CGA180M/CGA110F needle valve installed. The recommended regulators, control valves, and lecture-bottle stations are specified on the product landing page in the related products section. The recommended regulators suitable for corrosive gases are either Product Z148504(delivery pressure 0-15 psi) or Product Z148512(delivery pressure 0-50 psi).

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service