Skip to Content
Merck
All Photos(2)

Key Documents

290602

Sigma-Aldrich

Dipropyl phthalate

98%

Synonym(s):

Di-n-Propyl phthalate, Dipropyl benzene-1,2-dicarboxylate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H4-1,2-(CO2CH2CH2CH3)2
CAS Number:
Molecular Weight:
250.29
Beilstein:
2332522
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

refractive index

n20/D 1.497 (lit.)

bp

317.5 °C (lit.)

density

1.078 g/mL at 25 °C (lit.)

SMILES string

CCCOC(=O)c1ccccc1C(=O)OCCC

InChI

1S/C14H18O4/c1-3-9-17-13(15)11-7-5-6-8-12(11)14(16)18-10-4-2/h5-8H,3-4,9-10H2,1-2H3

InChI key

MQHNKCZKNAJROC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Degradation of dipropyl phthalate by lypolytic enzymes (fungal cutinase, yeast esterase) has been investigated.

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Repr. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

228.2 °F - closed cup

Flash Point(C)

109 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yang-Hoon Kim et al.
Archives of microbiology, 184(1), 25-31 (2005-08-02)
The efficiency of two lypolytic enzymes (fungal cutinase, yeast esterase) in the degradation of dipropyl phthalate (DPrP) was investigated. The DPrP-degradation rate of fungal cutinase was surprisingly high, i.e., almost 70% of the initial DPrP (500 mg/l) was decomposed within
Hung-Hung Sung et al.
Fish & shellfish immunology, 31(3), 511-515 (2011-07-12)
This study set out to understand the sublethal effect of xenobiotic phthalate esters (PAEs) on the relationship between the susceptibility of shrimp to bacterial infection and the immune response of the shrimp. Neocaridina denticulate were exposed to different concentrations of
Takashi Maruyama et al.
Microbiology and immunology, 51(3), 321-326 (2007-03-24)
Phthalate esters with short alkyl chains, such as di-ethyl (DEP), di-n-propyl (DPP), and di-butyl phthalate (DBP), have adjuvant effects on an FITC-induced contact hypersensitivity mouse model. The adjuvant effects of DPP and DBP are associated with enhanced trafficking of FITC-presenting
Y Imai et al.
Clinical and experimental allergy : journal of the British Society for Allergy and Clinical Immunology, 36(11), 1462-1468 (2006-11-07)
Many different types of phthalate ester are used as plasticizers and are thus found in the air. There have been several studies that suggest an association between allergies and phthalate esters. We previously found that di-butyl phthalate (DBP) has an
Reproductive toxicology. Di-n-propylphthalate.
Environmental health perspectives, 105 Suppl 1, 257-258 (1997-02-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service